Skip to Content
Merck
All Photos(2)

Key Documents

29185

Millipore

Vegetable Peptone for Biotechnology and Microbiology

Proteose Peptone (vegetable)

Proteose, Encymatically Digested, for Biotechnology and Microbiology

Synonym(s):

Peptone from vegetable, Peptone (vegetable), No. 3, Vegetable Proteose Peptone

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€137.00

€137.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
€137.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

€137.00


Estimated to ship on29 May 2025


Request a Bulk Order

Product Name

Proteose Peptone (vegetable), suitable for microbiology

biological source

plant

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

amino-nitrogen, ≥3.2%
total nitrogen (N), ≥11%

impurities

≤12% ash

loss

≤6% loss on drying

pH

6.6±0.5 (25 °C, 2% in H2O)

solubility

H2O: 2%, clear, yellow

application(s)

food and beverages
vitamins, nutraceuticals, and natural products

microbiology

Looking for similar products? Visit Product Comparison Guide

General description

Proteose Peptone (vegetable) is an enzymatic hydrolysate of vegetable protein and therefore an animal-free alternative to traditional peptones. It is recommended for the cultivation of fastidious pathogens and can be employed in media or bulk production of antibiotics, enzymes, veterinary preparation, bacterial toxins, etc.

Application

Proteose Peptone (vegetable) facilitates the growth of a broad range of microbes, from fastidious aerobic bacteria to yeasts and molds. It is recommended for use in media that support good growth of a large number of microorganisms including Staphylococci, Streptococci, Pneumococci, Meningococci, and Gonococci, among others which require a highly nutritious medium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Federico Rojas et al.
Cell, 176(1-2), 306-317 (2018-12-07)
Trypanosome parasites control their virulence and spread by using quorum sensing (QS) to generate transmissible "stumpy forms" in their host bloodstream. However, the QS signal "stumpy induction factor" (SIF) and its reception mechanism are unknown. Although trypanosomes lack G protein-coupled

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service