Skip to Content
Merck
All Photos(2)

Key Documents

96174

Millipore

Pea Peptone for Biotechnology and Microbiology

Enzymatically Digested, for Biotechnology and Microbiology

Synonym(s):

Peptone from vegetable

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€152.00

€152.00


Estimated to ship on01 June 2025


Request a Bulk Order

Select a Size

Change View
500 G
€152.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

€152.00


Estimated to ship on01 June 2025


Request a Bulk Order

Product Name

Peptone from pea, suitable for microbiology

biological source

plant

Quality Level

form

powder

composition

amino-nitrogen, ≥3.5%
total nitrogen (N), ≥13%

loss

≤6% loss on drying

pH

6.7-7.7

application(s)

food and beverages
microbiology

Looking for similar products? Visit Product Comparison Guide

Application

Peptone from pea is manufactured by a controlled enzymatic hydrolysis with raw materials from vegetal origin only. It contains a mix of peptides and free amino acids and is guaranteed "non GMO" according to the European Regulations 1829/2003 and 1830/2003.
Source of organic nitogen and growth factors recommended in media for industrial fermentation, cell culture and analytical microbiology.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debra Rossouw et al.
Applied and environmental microbiology, 75(20), 6600-6612 (2009-08-25)
Commercial wine yeast strains of the species Saccharomyces cerevisiae have been selected to satisfy many different, and sometimes highly specific, oenological requirements. As a consequence, more than 200 different strains with significantly diverging phenotypic traits are produced globally. This genetic

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service