Skip to Content
Merck
All Photos(1)

Key Documents

68971

Millipore

Casein-Meat Peptone for Biotechnology and Microbiology

Encymatically Digested, for Biotechnology and Microbiology, ≥12% Total N, ≥3.5% Amino

Synonym(s):

Peptone from bovine meat, Neopeptone, Special Peptone

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
€133.00

€133.00


Estimated to ship on30 May 2025


Request a Bulk Order

Select a Size

Change View
500 G
€133.00

About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
NACRES:
NA.85

€133.00


Estimated to ship on30 May 2025


Request a Bulk Order

Product Name

Peptone special, suitable for microbiology

biological source

bovine milk

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

composition

amino-nitrogen, ≥3.5%
total nitrogen (N), ≥12%

impurities

≤15% ash
≤5% NaCl
≤5% water

loss

≤11% loss on drying

pH

6.5-7.5 (2% in H2O)

solubility

H2O: 2%, clear, yellow

application(s)

food and beverages
microbiology

Looking for similar products? Visit Product Comparison Guide

General description

Peptone Special is an ingredient of media where a wide range of fastidious organisms are to be cultured. It is an enzymatic digest of animal tissues, carefully processed to ensure consistent growth characteristics when used in differential media.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Debra Rossouw et al.
Applied and environmental microbiology, 75(20), 6600-6612 (2009-08-25)
Commercial wine yeast strains of the species Saccharomyces cerevisiae have been selected to satisfy many different, and sometimes highly specific, oenological requirements. As a consequence, more than 200 different strains with significantly diverging phenotypic traits are produced globally. This genetic

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service