Skip to Content
Merck

Skip To

252425

Rhodamine B

Dye content 90 %

Synonym(s):

Basic Violet 10, Brilliant Pink B, Rhodamine O, Tetraethylrhodamine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C28H31ClN2O3
CAS Number:
Molecular Weight:
479.01
EC Number:
201-383-9
UNSPSC Code:
12171500
NACRES:
NA.47
PubChem Substance ID:
MDL number:
Colour Index Number:
45170
Beilstein/REAXYS Number:
4091619
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


composition

Dye content, 90%

mp

210-211 (dec.) (lit.)

λmax

543 nm

SMILES string

[Cl-].CCN(CC)c1ccc2c(OC3=CC(\C=CC3=C2c4ccccc4C(O)=O)=[N+](/CC)CC)c1

InChI

1S/C28H30N2O3.ClH/c1-5-29(6-2)19-13-15-23-25(17-19)33-26-18-20(30(7-3)8-4)14-16-24(26)27(23)21-11-9-10-12-22(21)28(31)32;/h9-18H,5-8H2,1-4H3;1H

InChI key

PYWVYCXTNDRMGF-UHFFFAOYSA-N

Application

Suitable as laser dye. Colorimetric reagent for Cd.

Features and Benefits

Powerful dye.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
0255879754R6626
composition

Dye content, 90%

composition

-

composition

-

composition

-

mp

210-211 (dec.) (lit.)

mp

-

mp

210-211 (dec.) (lit.)

mp

210-211 (dec.) (lit.)

λmax

543 nm

λmax

-

λmax

-

λmax

-


Still not finding the right product?

Explore all of our products under Rhodamine B


pictograms

CorrosionExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content


Van Duong Ta et al.
Scientific reports, 3, 1362-1362 (2013-03-02)
Optical microcavities are important for both fundamental studies of light-matter interaction and applications such as microlasers, optical switches and filters etc... Tunable microresonators, in which resonant modes can be manipulated, are especially fascinating. Here we demonstrate a unique approach to
Linda Johansson et al.
Ultrasonics, 53(5), 1020-1032 (2013-03-19)
An acoustic trap with miniaturized integrated transducers (MITs) for applications in non-contact trapping of cells or particles in a microfluidic channel was characterized by measuring the temperature increase and trapping strength. The fluid temperature was measured by the fluorescent response
Ewa Anna Blazejewska et al.
Stem cells (Dayton, Ohio), 27(3), 642-652 (2008-12-17)
The aim of this study was to investigate the transdifferentiation potential of murine vibrissa hair follicle (HF) stem cells into corneal epithelial-like cells through modulation by corneal- or limbus-specific microenvironmental factors. Adult epithelial stem cells were isolated from the HF



Questions

Reviews

No rating value

Active Filters