Skip to Content
Merck
All Photos(1)

Key Documents

24201-M

Sigma-Aldrich

Acetone

puriss., meets analytical specification of Ph. Eur., BP, NF, ≥99% (GC)

Sign Into View Organizational & Contract Pricing

Select a Size

1 L
€30.30
2.5 L
€84.90

€30.30


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 L
€30.30
2.5 L
€84.90

About This Item

Linear Formula:
CH3COCH3
CAS Number:
Molecular Weight:
58.08
Beilstein:
635680
EC Number:
MDL number:
UNSPSC Code:
12352115
eCl@ss:
39021201
PubChem Substance ID:
NACRES:
NA.02
technique(s):
GC/GC: suitable
bp:
56 °C/760 mmHg (lit.)
vapor pressure:
184 mmHg ( 20 °C)

€30.30


Estimated to ship on28 May 2025


Request a Bulk Order

vapor density

2 (vs air)

Quality Level

vapor pressure

184 mmHg ( 20 °C)

grade

puriss.

Assay

≥99% (GC)

form

liquid

quality

meets analytical specification of Ph. Eur., BP, NF

expl. lim.

13.2 %

technique(s)

GC/GC: suitable

impurities

acidity or alkalinity, complies
reducing matter, complies
related subst., complies (GC)
residual solvents, complies
water-insoluble matter, complies
≤0.0001% heavy metals (as Pb)
≤0.0002% benzene (GC)
≤0.002% non-volatile matter
≤0.05% 2-propanol (GC)
≤0.05% methanol (GC)
≤0.3% water (Karl Fischer)

refractive index

n20/D 1.359 (lit.)

bp

56 °C/760 mmHg (lit.)

mp

−94 °C (lit.)

density

0.790-0.792 g/mL at 20 °C
0.791 g/mL at 25 °C (lit.)

suitability

complies for appearance of solution
complies for identity (IR)

format

neat

SMILES string

CC(C)=O

InChI

1S/C3H6O/c1-3(2)4/h1-2H3

InChI key

CSCPPACGZOOCGX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetone is the most widely used solvent for cleaning purposes. It is also used as a precursor in the manufacture of polymers, bisphenol A, and methyl methacrylate.

Application

Acetone can be used as a solvent in the:
  • Ozonolysis of alkenes to aldehydes or ketones in the presence of solubilized water.
  • Catalyst-free glycerolysis of sunflower oil to synthesize partial glycerides.
  • Free radical polymerization of styrene to synthesize polystyrene.
  • Lipase-catalyzed esterification of isoascorbic acid with palmitic acid to form isoascorbyl palmitate.
Acetone′s luminesence intensity is dependent upon the solution components . The absorption of UV light by acetone, results in its photolysis and the production of radials .

Features and Benefits

  • Relatively low toxicity compared to other industrial solvents
  • Highly volatile
  • Ozone-stable solvent

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simplified spectrophotometric determination of ester groups in lipids.
F SNYDER et al.
Biochimica et biophysica acta, 34, 244-245 (1959-07-01)
Gianluigi Luppi et al.
The Journal of organic chemistry, 70(18), 7418-7421 (2005-08-27)
[reaction: see text] The aldol condensation of acetone with several isatins is described. The desired compound was obtained in quantitative yield and with good enantioselectivities up to 77%. The best results were obtained with 10 mol % H-D-Pro-L-beta3-hPhg-OBn as a
Gianluca Molla et al.
The Biochemical journal, 464(3), 387-399 (2014-10-01)
The aaoSo gene from Streptococcus oligofermentans encodes a 43 kDa flavoprotein, aminoacetone oxidase (SoAAO), which was reported to possess a low catalytic activity against several different L-amino acids; accordingly, it was classified as an L-amino acid oxidase. Subsequently, SoAAO was demonstrated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service