Skip to Content
Merck
All Photos(1)

Key Documents

21319-U

Supelco

Silica Gel

10% AgNO₃ Coated, pkg of 100 g bulk packing

Synonym(s):

Silver Nitrate Coated Silica Gel

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
13111000
NACRES:
NB.21
Pricing and availability is not currently available.

Product Name

10% AgNO3 Coated Silica Gel, pkg of 100 g bulk packing

Agency

suitable for EPA 1613

Quality Level

form

solid

packaging

pkg of 100 g bulk packing

technique(s)

solid phase extraction (SPE): suitable

particle size

60-200 mesh

application(s)

food and beverages

General description

The treated silica gels, whcih may be typically used in the pre-packed multi-layer silica gel columns, are offered in bulk packages. They can be useful for customizing the desired columns to efficiently clean very dirty samples, or to prepare shorter columns when the analytes are relatively clean, e.g. drinking water.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Repr. 1B - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Related Content

This page is intended to make it easier to find the consumables you need based on the analytical method you’re using. Methods included on this page come from the EPA, Standard Methods and ASTM.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service