8.22251
Acetone
EMPLURA®
Synonym(s):
Dimethyl ketone, Propanone, 2-Propanone
About This Item
Recommended Products
grade
for analytical purposes
for cleaning
for extraction
for preparative purposes
for synthesis
Quality Level
vapor density
2 (vs air)
vapor pressure
184 mmHg ( 20 °C)
product line
EMPLURA®
Assay
≥99% (GC)
form
liquid
autoignition temp.
465 °C (DIN 51794)
potency
5800 mg/kg LD50, oral (Rat)
20000 mg/kg LD50, skin (Rabbit)
expl. lim.
13.2 %
technique(s)
UV/Vis spectroscopy: suitable
impurities
≤0.3% Water
evapn. residue
≤0.0040%
refractive index
n20/D 1.359 (lit.)
pH
5-6 (20 °C, 395 g/L in H2O)
capacity
1000 g/kg water absorbing capacity
bp
56 °C/760 mmHg (lit.)
mp
−94 °C (lit.)
transition temp
flash point <-20 °C
density
0.791 g/mL at 25 °C (lit.)
application(s)
environmental
food and beverages
industrial qc
pharmaceutical
format
neat
storage temp.
2-30°C
SMILES string
CC(C)=O
InChI
1S/C3H6O/c1-3(2)4/h1-2H3
InChI key
CSCPPACGZOOCGX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Analysis Note
Density (d 20 °C/ 4 °C): 0.789 - 0.791
Evaporation residue: ≤ 0.0040 %
Water: ≤ 0.30 %
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3
Target Organs
Central nervous system
Supplementary Hazards
Storage Class Code
3 - Flammable liquids
WGK
WGK 1
Flash Point(F)
1.4 °F - closed cup
Flash Point(C)
-17.0 °C - closed cup
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Related Content
Our ergonomic 1 L HDPE bottle for acids, bases, and solvents provide an alternative to glass chemical bottles for lab safety in pouring & handling of hazardous chemicals in the laboratory.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service