Skip to Content
Merck

Skip To

08503

Fmoc-D-Cys(Trt)-OH

≥97.0% (HPLC)

Synonym(s):

Nα-Fmoc-S-trityl-D-cysteine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
5 g
Check Cart for Availability
€ 114,00

About This Item

Empirical Formula (Hill Notation):
C37H31NO4S
CAS Number:
Molecular Weight:
585.71
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26
MDL number:

€ 114,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97.0% (HPLC)

form

powder or crystals

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

color

white

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

OC(=O)[C@@H](CSC(c1ccccc1)(c2ccccc2)c3ccccc3)NC(=O)OCC4c5ccccc5-c6ccccc46

InChI

1S/C37H31NO4S/c39-35(40)34(38-36(41)42-24-33-31-22-12-10-20-29(31)30-21-11-13-23-32(30)33)25-43-37(26-14-4-1-5-15-26,27-16-6-2-7-17-27)28-18-8-3-9-19-28/h1-23,33-34H,24-25H2,(H,38,41)(H,39,40)/t34-/m1/s1

InChI key

KLBPUVPNPAJWHZ-UUWRZZSWSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
8.5234845931347695
form

powder or crystals

form

powder

form

-

form

-

assay

≥97.0% (HPLC)

assay

≥95% (TLC), ≥97.0% (HPLC)

assay

97%

assay

≥95.0% (sum of enantiomers, HPLC)

functional group

Fmoc

functional group

thiol

functional group

-

functional group

Fmoc

storage temp.

2-8°C

storage temp.

15-25°C

storage temp.

-

storage temp.

2-8°C

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

100

color

white

color

-

color

-

color

-


Still not finding the right product?

Explore all of our products under Fmoc-D-Cys(Trt)-OH


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
281662-2L04061826225646
281662-1L04061826225639
802934250004022536377216
281662-100ML04061826225622
04143-1ML-F04061838629159
04143-5ML-F04061836957247
M14102-500ML04061834040620
M14102-4L04061837230011
281662-PZ04061823900799
802934025004022536377193

Questions

Reviews

No rating value

Active Filters