Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
50 discs | Check Cart for Availability | € 26,40 |
About This Item
product line
BioChemika
Quality Segment
shelf life
limited shelf life, expiry date on the label
technique(s)
microbe id | susceptibility testing: suitable
application(s)
clinical testing
environmental
food and beverages, microbiology
shipped in
wet ice
storage temp.
2-8°C
suitability
Streptococcus spp.
General description
Packaging
1 of 1
This Item | |||
|---|---|---|---|
| product line BioChemika | product line BioChemika | product line BioChemika | product line BioChemika |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C | storage temp. 2-8°C |
| suitability Streptococcus spp. | suitability Streptococcus spp. | suitability Citrobacter spp., Enterobacter spp., Escherichia coli, Klebsiella spp., Proteus spp., Salmonella spp., Serratia spp., Shigella spp., Streptococcus spp., bacteria spp. | suitability Streptococcus spp., bacteria |
| technique(s) microbe id | susceptibility testing: suitable | technique(s) microbe id | specific enzyme detection: suitable | technique(s) microbe id | utilization test: suitable | technique(s) microbe id | utilization test: suitable |
| shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label | shelf life limited shelf life, expiry date on the label |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
