Skip to Content
Merck

Skip To

06985

Methyl 3-aminobenzoate

≥97.0% (GC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
10 g
Check Cart for Availability
€ 217,00
€ 162,75

About This Item

Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16
UNSPSC Code:
12352209
NACRES:
NA.22
PubChem Substance ID:
EC Number:
224-842-5
Beilstein/REAXYS Number:
1101468
MDL number:

€ 162,75

List Price€ 217,00Save 25%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97.0% (GC)

form

crystals or chunks

reaction suitability

reaction type: solution phase peptide synthesis

color

white to brown

mp

≥42 °C

application(s)

peptide synthesis

SMILES string

COC(=O)c1cccc(N)c1

InChI

1S/C8H9NO2/c1-11-8(10)6-3-2-4-7(9)5-6/h2-5H,9H2,1H3

InChI key

VZDNXXPBYLGWOS-UHFFFAOYSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
E10505PHR9177PHR9173
reaction suitability

reaction type: solution phase peptide synthesis

reaction suitability

-

reaction suitability

-

reaction suitability

-

Quality Level

100

Quality Level

100

Quality Level

300

Quality Level

300

assay

≥97.0% (GC)

assay

97%

assay

-

assay

-

mp

≥42 °C

mp

-

mp

110-111 °C (lit.)

mp

-

application(s)

peptide synthesis

application(s)

-

application(s)

pharmaceutical small molecule

application(s)

pharmaceutical small molecule

form

crystals or chunks

form

-

form

-

form

-


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters