Skip to Content
Merck

Skip To

02451

Fmoc-N-Me-D-Leu-OH

≥97.0%

Synonym(s):

Fmoc-N-methyl-D-leucine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 g
Check Cart for Availability
€ 114,00
€ 85,50

About This Item

Empirical Formula (Hill Notation):
C22H25NO4
CAS Number:
Molecular Weight:
367.44
UNSPSC Code:
12352209
NACRES:
NA.26
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
7350417

€ 85,50

List Price€ 114,00Save 25%
Web-Only Promotion

Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥97.0%

form

powder

reaction suitability

reaction type: Fmoc solid-phase peptide synthesis

color

colorless to white

mp

108-116 °C

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

CC(C)C[C@@H](N(C)C(=O)OCC1c2ccccc2-c3ccccc13)C(O)=O

InChI

1S/C22H25NO4/c1-14(2)12-20(21(24)25)23(3)22(26)27-13-19-17-10-6-4-8-15(17)16-9-5-7-11-18(16)19/h4-11,14,19-20H,12-13H2,1-3H3,(H,24,25)/t20-/m1/s1

InChI key

BUJQSIPFDWLNDC-HXUWFJFHSA-N


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
023994763347316
application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

application(s)

peptide synthesis

form

powder

form

powder or crystals

form

-

form

-

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

assay

≥97.0%

assay

≥98.0% (HPLC)

assay

≥97.0%

assay

≥95.0% (TLC)

functional group

Fmoc

functional group

Fmoc

functional group

Fmoc, amine, carboxylic acid

functional group

Fmoc


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters