Skip to Content
Merck
All Photos(1)

Key Documents

40-4001

Sigma-Aldrich

Methyl orange-xylene cyanole FF solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000

form

liquid

availability

available only in Japan

Suitability

for titration

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service