Skip to Content
Merck
All Photos(1)

Key Documents

1.01322

Supelco

Methyl orange (C.I. 13025)

pH indicator, ACS, Reag. Ph Eur

Synonym(s):

Methyl orange (C.I. 13025), 4-Dimethylaminoazobenzene-4′-sulfonic acid sodium salt, Gold orange, Helianthine, Orange III

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
€74.00
100 G
€259.00
1 KG
€1,850.00

€74.00


Estimated to ship on27 March 2025


Request a Bulk Order

Select a Size

Change View
25 G
€74.00
100 G
€259.00
1 KG
€1,850.00

About This Item

Empirical Formula (Hill Notation):
C14H14N3NaO3S
CAS Number:
Molecular Weight:
327.33
MDL number:
UNSPSC Code:
12352107
EC Index Number:
208-925-3
NACRES:
NA.05
grade:
ACS reagent

€74.00


Estimated to ship on27 March 2025


Request a Bulk Order

Product Name

Methyl orange (C.I. 13025), indicator ACS,Reag. Ph Eur

grade

ACS reagent

Quality Level

Agency

reag. Ph. Eur.

form

solid

potency

60 mg/kg LD50, oral (Rat)

loss

≤5% loss on drying, 110°C

pH

6.5 (20 °C, 5 g/L in H2O)

mp

>300 °C

bulk density

200‑400 kg/m3

storage temp.

2-30°C

SMILES string

[S](=O)(=O)([O-])c1ccc(cc1)N=Nc2ccc(cc2)N(C)C

InChI

1S/C14H15N3O3S/c1-17(2)13-7-3-11(4-8-13)15-16-12-5-9-14(10-6-12)21(18,19)20/h3-10H,1-2H3,(H,18,19,20)/p-1

InChI key

IETWCRRCPURZOC-UHFFFAOYSA-M

Application

  • Plant-assisted green preparation of silver nanoparticles using leaf extract of Dalbergia sissoo and their antioxidant, antibacterial and catalytic applications.: This research utilized Methyl Orange as an indicator in the green synthesis of silver nanoparticles using Dalbergia sissoo leaf extract. The study highlighted its catalytic application in dye degradation, demonstrating the environmental benefits of plant-assisted nanoparticle synthesis (Khatun et al., 2024).
  • Dye Degradation and Sulfur Oxidation of Methyl Orange and Thiophenol via Newly Designed Nanocomposite GQDs/NiSe-NiO Photocatalyst Under Homemade LED Light.: This study focused on the efficient photocatalytic degradation of Methyl Orange using a novel nanocomposite photocatalyst under LED light, presenting significant advancements in photocatalytic materials for wastewater treatment (Srivastava et al., 2023).
  • Bryophyllum pinnatum leaf extract mediated ZnO nanoparticles with prodigious potential for solar driven photocatalytic degradation of industrial contaminants.: The research demonstrated the synthesis of ZnO nanoparticles using Bryophyllum pinnatum leaf extract and their application in the photocatalytic degradation of Methyl Orange, showcasing a sustainable approach to tackling industrial pollutants (Dhiman et al., 2023).
  • Light driven Aspergillus niger-ZnS nanobiohybrids for degradation of methyl orange.: This paper explored the use of Aspergillus niger-ZnS nanobiohybrids for the degradation of Methyl Orange, emphasizing the potential of biogenic approaches in enhancing photocatalytic efficiency (Priyanka & Lens, 2022).
  • Bacterially driven cadmium sulfide precipitation on porous membranes: Toward platforms for photocatalytic applications.: This study utilized Methyl Orange to assess the photocatalytic efficiency of cadmium sulfide precipitated on porous membranes by bacteria, proposing a novel method for creating photocatalytic platforms for environmental applications (Marusak et al., 2018).

Analysis Note

Transition range: pH 3.1 - pH 4.4pink - orange yellow
Appearance of solution: passes test
Loss on drying (110 °C): ≤ 5
Transition range (according to ACS): passes test
Sensitivity test: passes test

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible, acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service