Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Linear Formula:
Ag2CO3
CAS Number:
Molecular Weight:
275.75
PubChem Substance ID:
UNSPSC Code:
12352300
Beilstein/REAXYS Number:
6936654
MDL number:
Assay:
≥98.0%
Grade:
SAJ first grade
Form:
solid
Pricing and availability is not currently available.
grade
SAJ first grade
assay
≥98.0%
form
solid
availability
available only in Japan
mp
210 °C (dec.) (lit.)
density
6.08 g/mL at 25 °C (lit.)
storage temp.
15-25°C
SMILES string
[Ag]OC(=O)O[Ag]
InChI
1S/CH2O3.2Ag/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2
InChI key
KQTXIZHBFFWWFW-UHFFFAOYSA-L
Still not finding the right product?
Explore all of our products under Silver carbonate
signalword
Danger
hcodes
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1
Storage Class
13 - Non Combustible Solids
wgk
WGK 2
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Roman V Rozhkov et al.
The Journal of organic chemistry, 68(16), 6314-6320 (2003-08-05)
The palladium-catalyzed annulation of 1,3-dienes by o-iodoacetoxycoumarins provides an efficient method for the synthesis of biologically interesting dihydrofurocoumarins. The presence of the acetyl group on the phenolic oxygen and the use of silver carbonate as a base are crucial for
José M López-Cepero
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 52(2), 211-216 (2004-01-20)
Silver staining methods, when selective, yield a high-contrast and high-resolution image in optical microscopy. A classical method for silver impregnation of mitochondria has been applied to murine tissues and reveals a marked heterogeneity among mitochondria in single cells. This heterogeneity
Palladium-catalyzed decarboxylative C-H bond arylation of thiophenes.
Peng Hu et al.
Angewandte Chemie (International ed. in English), 51(1), 227-231 (2011-11-17)

