Skip to Content
Merck
All Photos(1)

Key Documents

24-4820

Sigma-Aldrich

Potassium iodate

SAJ first grade, ≥99.5%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KIO3
CAS Number:
Molecular Weight:
214.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99.5%
grade:
SAJ first grade
form:
crystalline
Pricing and availability is not currently available.

grade

SAJ first grade

Assay

≥99.5%

form

crystalline

availability

available only in Japan

mp

560 °C (lit.)

density

3.93 g/mL at 25 °C (lit.)

SMILES string

[K+].[O-]I(=O)=O

InChI

1S/HIO3.K/c2-1(3)4;/h(H,2,3,4);/q;+1/p-1

InChI key

JLKDVMWYMMLWTI-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ralf Biebinger et al.
The British journal of nutrition, 102(9), 1362-1369 (2009-08-06)
Adverse sensory changes prevent the addition of highly bioavailable ferrous sulfate (FeSO4) to most wheat flours. Poorly absorbable reduced Fe powders are commonly used. Encapsulation of FeSO4 can overcome these sensory changes, but the particle size of commercial compounds is
Fabian Rohner et al.
Food and nutrition bulletin, 33(4 Suppl), S330-S335 (2013-03-01)
Despite considerable progress made in the past decade through salt iodization programs, over 2 billion people worldwide still have inadequate iodine intake, with devastating consequences for brain development and intellectual capacity. To optimize these programs with regard to salt iodine
Wim Voogt et al.
Journal of the science of food and agriculture, 90(5), 906-913 (2010-04-01)
Iodine is an essential trace element for humans. Two billion individuals have insufficient iodine intake. Biofortification of vegetables with iodine offers an excellent opportunity to increase iodine intake by humans. The main aim was to study the effect of iodine
Gehad G Mohamed et al.
Drug testing and analysis, 2(1), 28-36 (2010-09-30)
Two spectrophotometric procedures are suggested for the determination of three irreversible proton pump inhibitors, rabeprazole (RAB), omeprazole (OMP) and pantoprazole (PAN) in pure form and in different pharmaceutical formulations. The first method is based on the oxidation of RAB and
Xue F Yang et al.
The British journal of nutrition, 98(1), 116-122 (2007-04-26)
Excessive iodine induces thyroid dysfunction. However, the effect of excessive iodine exposure on maternal-fetal thyroid hormone metabolism and on the expression of genes involved in differentiation, growth and development is poorly understood. Since a thyroid hormone receptor response element was

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service