Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Pricing and availability is not currently available.
form
liquid
availability
available only in Japan
SMILES string
[Cu+2].[Na+].[Na+].[S](=O)(=O)(O)O.[O-]C(=O)C(O)(CC(=O)[O-])CC(=O)[O-].[O-]C(=O)O.O
InChI
1S/C6H8O7.CH2O3.Cu.2Na.H2O4S.H2O/c7-3(8)1-6(13,5(11)12)2-4(9)10;2-1(3)4;;;;1-5(2,3)4;/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12);(H2,2,3,4);;;;(H2,1,2,3,4);1H2/q;;+2;2*+1;;/p-4
InChI key
QFKWSRIUZIYLCK-UHFFFAOYSA-J
Still not finding the right product?
Explore all of our products under Benedict′s reagent
signalword
Warning
hcodes
Hazard Classifications
Aquatic Chronic 2 - Eye Irrit. 2
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

