Skip to Content
Merck
All Photos(1)

Key Documents

01-1400

Sigma-Aldrich

Activated carbon

SAJ first grade

Synonym(s):

Activated charcoal, Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

vapor pressure

<0.1 mmHg ( 20 °C)

form

powder or granules

autoignition temp.

842 °F

availability

available only in Japan

resistivity

1375 μΩ-cm, 20°C (graphite)

mp

3550 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elisangela Heiderscheidt et al.
Journal of environmental sciences (China), 25(4), 659-669 (2013-08-09)
The drainage of peatland areas for peat extraction, agriculture or bioenergy requires affordable, simple and reliable treatment methods that can purify waters rich in particulates and dissolved organic carbon. This work focused on the optimisation of chemical purification process for
T Southorn et al.
Journal of perioperative practice, 23(6), 144-146 (2013-08-06)
Currently, there are very few provisions for recycling in theatres. We measured the weight of clinical waste for several orthopaedic operations. This waste was then examined and sorted into domestic waste and clinical waste. With staff education it is possible
Guofeng Shen et al.
Journal of environmental sciences (China), 25(3), 511-519 (2013-08-08)
The uncertainty in emission estimation is strongly associated with the variation in emission factor (EF), which could be influenced by a variety of factors such as fuel properties, stove type, fire management and even methods used in measurements. The impacts
Christopher M Gough et al.
Ecological applications : a publication of the Ecological Society of America, 23(5), 1202-1215 (2013-08-24)
Carbon (C) uptake rates in many forests are sustained, or decline only briefly, following disturbances that partially defoliate the canopy. The mechanisms supporting such functional resistance to moderate forest disturbance are largely unknown. We used a large-scale experiment, in which
Christopher D Cappa et al.
Science (New York, N.Y.), 339(6118), 393-393 (2013-09-04)
Jacobson argues that our statement that "many climate models may overestimate warming by BC" has not been demonstrated. Jacobson challenges our results on the basis that we have misinterpreted some model results, omitted optical focusing under high relative humidity conditions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service