Skip to Content
Merck
All Photos(1)

Key Documents

NIST2451

Fine carbon (activated)

NIST® SRM® 2451, from cyanide ore leaching

Synonym(s):

Carbon

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€1,420.00

€1,420.00


Available to ship on16 April 2025Details


Request a Bulk Order

Select a Size

Change View
100 G
€1,420.00

About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
UNSPSC Code:
41116107
NACRES:
NA.24

€1,420.00


Available to ship on16 April 2025Details


Request a Bulk Order

grade

certified reference material

Quality Level

packaging

pkg of 100 g

manufacturer/tradename

NIST®

application(s)

general analytical

format

matrix material

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

The SRM 2451 unit contains a bottle with 100 g of activated fine-powder carbon, which is obtained through the leaching of ore with a cyanide solution. For more information, please refer to the SDS and COA.
SRM 2451_cert

SRM 2451_SDS

Application

Fine carbon standard referencematerial (SRM) is intended to evaluate the chemical analysis methods and tocalibrate the instrumental methods of analysis.

Features and Benefits

  • NIST certified values for mercury are provided, values are metrologically traceable to the SI unit of mass, expressed as milligrams per kilogram.
  • A reference value for gold is reported based on dry mass basis and determined by inductively coupled plasma massspectrometry (ICP-MS).
  • The information value for cyanide is provided and is determined using EPA method SW-846 9012a, and the mass fraction is reported on a dry mass basis.
  • Expiration details and instructions for use are provided on the NIST certificate.

Other Notes

Certified for the analytes listed below.
Mercury (Hg)

Reference/Informational value is provided for the following analytes.
Gold (Au), Cyanide (CN)

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service