Merck
  • Optimisation of chemical purification conditions for direct application of solid metal salt coagulants: treatment of peatland-derived diffuse runoff.
Optimisation of chemical purification conditions for direct application of solid metal salt coagulants: treatment of peatland-derived diffuse runoff.
Journal of environmental sciences (China) (2013-08-09)
Elisangela Heiderscheidt, Jaakko Saukkoriipi, Anna-Kaisa Ronkanen, Bjørn Kløve
ABSTRACT
The drainage of peatland areas for peat extraction, agriculture or bioenergy requires affordable, simple and reliable treatment methods that can purify waters rich in particulates and dissolved organic carbon. This work focused on the optimisation of chemical purification process for the direct dosage of solid metal salt coagulants. It investigated process requirements of solid coagulants and the influence of water quality, temperature and process parameters on their performance. This is the first attempt to provide information on specific process requirements of solid coagulants. Three solid inorganic coagulants were evaluated: aluminium sulphate, ferric sulphate and ferric aluminium sulphate. Pre-dissolved aluminium and ferric sulphate were also tested with the objective of identifying the effects of in-line coagulant dissolution on purification performance. It was determined that the pre-dissolution of the coagulants had a significant effect on coagulant performance and process requirements. Highest purification levels achieved by solid coagulants, even at 30% higher dosages, were generally lower (5%-30%) than those achieved by pre-dissolved coagulants. Furthermore, the mixing requirements of coagulants pre-dissolved prior to addition differed substantially from those of solid coagulants. The pH of the water samples being purified had a major influence on coagulant dosage and purification efficiency. Ferric sulphate (70 mg/L) was found to be the best performing solid coagulant achieving the following load removals: suspended solids (59%-88%), total organic carbon (56%-62%), total phosphorus (87%-90%), phosphate phosphorus (85%-92%) and total nitrogen (33%-44%). The results show that the use of solid coagulants is a viable option for the treatment of peatland-derived runoff water if solid coagulant-specific process requirements, such as mixing and settling time, are considered.
Sigma-Aldrich
Activated Charcoal Norit®, Norit® CA1, wood, chemically activated, powder
Sigma-Aldrich
Activated Charcoal Norit®, Norit® GAC 1240W, from coal, for potable water processing, steam activated, granular
Sigma-Aldrich
Activated Charcoal Norit®, Norit® RB3, for gas purification, steam activated, rod
Sigma-Aldrich
Activated Charcoal Norit®, Norit® SX2, powder, from peat, multi-purpose activated charcoal, steam activated and acid washed
Sigma-Aldrich
Activated Charcoal Norit®, Norit® PK 1-3, from peat, steam activated, granular
Supelco
Activated charcoal, for the determination of AOX, 50-150 μm particle size
Sigma-Aldrich
Activated charcoal, suitable for cell culture, suitable for plant cell culture
Supelco
Activated Charcoal Norit®, Norit® RBAA-3, rod
Supelco
Activated charcoal, powder
Sigma-Aldrich
Carbon, glassy, spherical powder, 2-12 μm, 99.95% trace metals basis
Supelco
Activated charcoal, puriss. p.a., powder
Millipore
Activated charcoal, suitable for GC
Sigma-Aldrich
Carbon, nanopowder, <100 nm particle size (TEM)
Sigma-Aldrich
Activated charcoal, powder, -100 particle size (mesh), decolorizing
Sigma-Aldrich
Activated charcoal, acid-washed with hydrochloric acid
Sigma-Aldrich
Activated charcoal, untreated, granular, ≤5 mm
Sigma-Aldrich
Activated Charcoal, meets USP testing specifications
Sigma-Aldrich
Activated charcoal, untreated, granular, 8-20 mesh
Sigma-Aldrich
Activated charcoal, untreated, granular, 20-60 mesh
Sigma-Aldrich
Activated charcoal, DARCO®, 20-40 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 12-20 mesh, granular
Sigma-Aldrich
Activated charcoal, DARCO®, 4-12 mesh particle size, granular
Sigma-Aldrich
Activated charcoal, DARCO®, −100 mesh particle size, powder