Skip to Content
Merck
All Photos(1)

Key Documents

ATPD-RO

Roche

Adenosine 5′-triphosphate disodium salt trihydrate

Disodium salt

Synonym(s):

ATP

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€85.70
10 G
€132.00

€85.70


Estimated to ship on12 April 2025



Select a Size

Change View
5 G
€85.70
10 G
€132.00

About This Item

Empirical Formula (Hill Notation):
C10H20N5Na2O16P3
CAS Number:
Molecular Weight:
605.19
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:

€85.70


Estimated to ship on12 April 2025


Quality Level

Assay

≥98%

form

crystals

mol wt

Mr 507.2 (ATP)
Mr 605.2 (ATP-Na2 x 3 H2O)

packaging

pkg of 10 g (10127531001)
pkg of 5 g (10127523001)

manufacturer/tradename

Roche

storage temp.

2-8°C

SMILES string

O.O.O.[Na+].[Na+].Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O

InChI

1S/C10H16N5O13P3.2Na.3H2O/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(26-10)1-25-30(21,22)28-31(23,24)27-29(18,19)20;;;;;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H,23,24)(H2,11,12,13)(H2,18,19,20);;;3*1H2/q;2*+1;;;/p-2/t4-,6-,7-,10-;;;;;/m1...../s1

InChI key

MWEQTWJABOLLOS-AZGWGOJFSA-L

Looking for similar products? Visit Product Comparison Guide

General description

The product contains disodium salt crystals of ATP.

Application

ATP, disodium salt, is used for the luminometric determination of Luc activity in cell extracts.[1][2]

Quality

Purity: ≥98% ATP-Na2H2 x 3 H2O, ≥82% ATP (enzymatic), ≤0.5% ADP and AMP (enzymatic), ≤0.01% GTP (HPLC), ≤30 ppm heavy metals (as Pb-ions), ≤15 ppm Fe-ions

Preparation Note

Storage conditions (working solution): The reconstituted aqueous solution is stable for one week at 2 to 8 °C, and for several months at -15 to -25 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conazole fungicides inhibit Leydig cell testosterone secretion and androgen receptor activation in vitro
Roelofs
Toxicology, 271-283 (2014)
Direct chemiluminescence detection of nitric oxide in aqueous solutions using the natural nitric oxide target soluble guanylyl cyclase
Woldman
Free Radical Biology & Medicine, 47.10, 1339-1345 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service