Skip to Content
Merck
All Photos(1)

Key Documents

797189

Sigma-Aldrich

Adenosine 5′-triphosphate disodium salt trihydrate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
€62.90

€62.90


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
€62.90

About This Item

Empirical Formula (Hill Notation):
C10H20N5Na2O16P3
CAS Number:
Molecular Weight:
605.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€62.90


Check Cart for Availability

Request a Bulk Order

form

solid

functional group

ether
phosphate

SMILES string

O.O.O.[Na+].[Na+].Nc1ncnc2n(cnc12)[C@@H]3O[C@H](COP(O)(=O)OP([O-])(=O)OP(O)([O-])=O)[C@@H](O)[C@H]3O

InChI

1S/C10H16N5O13P3.2Na.3H2O/c11-8-5-9(13-2-12-8)15(3-14-5)10-7(17)6(16)4(26-10)1-25-30(21,22)28-31(23,24)27-29(18,19)20;;;;;/h2-4,6-7,10,16-17H,1H2,(H,21,22)(H,23,24)(H2,11,12,13)(H2,18,19,20);;;3*1H2/q;2*+1;;;/p-2/t4-,6-,7-,10-;;;;;/m1...../s1

InChI key

MWEQTWJABOLLOS-AZGWGOJFSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silke Lehmann et al.
PloS one, 15(6), e0234154-e0234154 (2020-06-05)
Phytohormones mediate most diverse processes in plants, ranging from organ development to immune responses. Receptor protein complexes perceive changes in intracellular phytohormone levels and trigger a signaling cascade to effectuate downstream responses. The in planta analysis of elements involved in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service