Skip to Content
Merck
All Photos(1)

Key Documents

1.30195

Millipore

HY-LiTE® ATP Standard Solution

Positive Control (PC) standard solution (1.0 ng/mL) for performance verification of HY-LiTE® 2/2A instruments, pkg of 5 vials (liquid standard solution)

Synonym(s):

ATP Standards, ATP positive control

Sign Into View Organizational & Contract Pricing

Select a Size

5 UNITS
€112.00

€112.00


Estimated to ship on30 May 2025


Request more information

Select a Size

Change View
5 UNITS
€112.00

About This Item

UNSPSC Code:
41112601
NACRES:
NA.24

€112.00


Estimated to ship on30 May 2025


Request more information

material

tube

product line

HY-LiTE®

form

liquid

feature

standard type

packaging

pkg of 5 vials (liquid standard solution)

manufacturer/tradename

HY-LiTE®

concentration

0.95-1.05 ng/mL

pH

11.7 (20 °C in H2O)

suitability

suitable for luminescence

application(s)

food and beverages
pharmaceutical

compatibility

for use with HY-LITE® instrument

detection method

bioluminescent

storage temp.

2-8°C

General description

The ATP Standard contains dissolved adenosine tri-phosphate. This can serve as a positive control for performance verification of HY-LiTE® 2/2A luminometers. Each tube contains approximately 1.4 ml ATP solution in a concentration of 1 ng/ml. Unopened tubes can be stored at room temperature until the expiry date printed on the pack. Once opened, each tube can be used once (maximally twice).

Application

The HY-LiTE® ATP Standard solution (1.0 ng/mL) is intended for performance verification of HY-LiTE® 2/2A luminometers.

Analysis Note

Appearance: clear, colourless
pH-value (25 °C): 11.00 - 12.00
Assay: 0.95 - 1.05 ng/ml

Legal Information

HY-LITE is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service