Skip to Content
Merck

Skip To

TX1278

Acetonitrile solution

suitable for HPLC, contains 0.05% (v/v) trifluoroacetic acid

Synonym(s):

Acetonitrile solution, ACN, Cyanomethane, Ethyl nitrile, Methyl cyanide, Trifluoroacetic Acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3CN
CAS Number:
Molecular Weight:
41.05
EC Index Number:
200-835-2
UNSPSC Code:
41116004
MDL number:
Beilstein/REAXYS Number:
4124158
Technique(s):
HPLC: suitable
Bp:
81.6 °C/1,013 hPa
Vapor pressure:
97 hPa ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Acetonitrile/TFA 0.05% Mobile Phase,

Quality Segment

vapor pressure

97 hPa ( 20 °C)

form

liquid

concentration

0.045-0.055 % (v/v) in trifluoroacetic acid

technique(s)

HPLC: suitable

evapn. residue

≤5 ppm

color

APHA: ≤10

bp

81.6 °C/1,013 hPa

mp

-45.7 °C

transition temp

flash point 2 °C

density

0.782 g/mL at 25 °C

UV absorption

λ: 190 nm Amax: ≤1.00, λ: 200 nm Amax: ≤0.50, λ: 210 nm Amax: ≤0.40, λ: 230 nm Amax: ≤0.30, λ: 250 nm Amax: ≤0.05, λ: 280 nm Amax: ≤0.010, λ: 300 nm Amax: ≤0.010, λ: 350 nm Amax: ≤0.010

format

mixture

shipped in

ambient

storage temp.

room temp

SMILES string

CC#N

InChI

1S/C2H3N/c1-2-3/h1H3

InChI key

WEVYAHXRMPXWCK-UHFFFAOYSA-N

Application

TX1278P-1 is packaged in poly coated glass bottles

Preparation Note

Product filtered through a 0.2 μm filter

Analysis Note

Assay (trifluoroacetic acid)0.045-0.055% (v/v)Color (APHA)10 maxEvaporation residue5ppm maxFiltered through 0.2 µm filterTo pass testFormClear liquidGradient at 210 nm0.005AU maxGradient at 220 nm0.001AU maxGradient at 280 nm0.001AU maxUV Abs. at 190 nm1.00AU maxUV Abs. at 200 nm0.50AU maxUV Abs. at 210 nm0.40AU maxUV Abs. at 230 nm0.30AU maxUV Abs. at 250 nm0.05AU maxUV Abs. at 280 nm0.010AU maxUV Abs. at 300 nm0.010AU maxUV Abs. at 350 nm0.010AU max


Still not finding the right product?


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters