Skip to Content
Merck

Skip To

SNAP2RL

SNAP id® 2.0 Blot Roller

New Blot Roller specially designed for use with the SNAP i.d. 2.0 system.

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
41116010
NACRES:
NB.22
eCl@ss:
42029053
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

manufacturer/tradename

SNAP id®

technique(s)

western blot: suitable

compatibility

for use with Commercially available blocking reagents, for use with Luminata Western HRP Substrates, for use with Nitrocellulose, for use with PVDF (Immobilon membranes), for use with blØk<TMSYMBOL></TMSYMBOL>-CH Buffer (cat. no. WBAVDCH01), for use with blØk<TMSYMBOL></TMSYMBOL>-FL Buffer (cat. no. WBAVDFL01), for use with blØk<TMSYMBOL></TMSYMBOL>-PO Buffer (cat. no. WBAVDP001), for use with commercially available detection reagents

detection method

chemiluminescent, colorimetric, fluorometric

shipped in

ambient

storage temp.

room temp

Application

New Blot Roller specially designed for use with the SNAP i.d. 2.0 system.

Legal Information

SNAP id is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SNAP2FRMN01SNAP2FRMDSNAP2BHMD0100
compatibility

for use with Commercially available blocking reagents, for use with Nitrocellulose, for use with blØk<TMSYMBOL></TMSYMBOL>-CH Buffer (cat. no. WBAVDCH01), for use with blØk<TMSYMBOL></TMSYMBOL>-PO Buffer (cat. no. WBAVDP001), for use with Luminata Western HRP Substrates, for use with blØk<TMSYMBOL></TMSYMBOL>-FL Buffer (cat. no. WBAVDFL01), for use with PVDF (Immobilon membranes), for use with commercially available detection reagents

compatibility

for use with Commercially available blocking reagents, for use with Luminata Western HRP Substrates, for use with Nitrocellulose, for use with PVDF (Immobilon membranes), for use with blØk<TMSYMBOL></TMSYMBOL>-CH Buffer (cat. no. WBAVDCH01), for use with blØk<TMSYMBOL></TMSYMBOL>-FL Buffer (cat. no. WBAVDFL01), for use with blØk<TMSYMBOL></TMSYMBOL>-PO Buffer (cat. no. WBAVDP001), for use with commercially available detection reagents

compatibility

for use with Commercially available blocking reagents, for use with Luminata Western HRP Substrates, for use with Nitrocellulose, for use with PVDF (Immobilon membranes), for use with blØk<TMSYMBOL></TMSYMBOL>-CH Buffer (cat. no. WBAVDCH01), for use with blØk<TMSYMBOL></TMSYMBOL>-FL Buffer (cat. no. WBAVDFL01), for use with blØk<TMSYMBOL></TMSYMBOL>-PO Buffer (cat. no. WBAVDP001), for use with commercially available detection reagents

compatibility

for use with Commercially available blocking reagents, for use with Luminata Western HRP Substrates, for use with Nitrocellulose, for use with PVDF (Immobilon membranes), for use with blØk<TMSYMBOL></TMSYMBOL>-CH Buffer (cat. no. WBAVDCH01), for use with blØk<TMSYMBOL></TMSYMBOL>-FL Buffer (cat. no. WBAVDFL01), for use with blØk<TMSYMBOL></TMSYMBOL>-PO Buffer (cat. no. WBAVDP001), for use with commercially available detection reagents

storage temp.

room temp

storage temp.

room temp

storage temp.

-

storage temp.

room temp

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient

detection method

chemiluminescent, fluorometric, colorimetric

detection method

chemiluminescent, colorimetric, fluorometric

detection method

chemiluminescent, colorimetric, fluorometric

detection method

chemiluminescent, colorimetric, fluorometric

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

technique(s)

western blot: suitable

technique(s)

western blot: suitable

technique(s)

western blot: suitable

technique(s)

western blot: suitable




Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Protocols

The SNAP i.d. 2.0 Protein Detection System is the second generation of the SNAP i.d. method for detecting immunoreactive proteins on Western blots.

Western blotting involves protein separation by gel electrophoresis, protein transfer to a polyvinylidene difluoride (PVDF) or nitrocellulose membrane, and detection by various methods.

Western-Blot-Protokoll mit Workflow-Schritten für verschiedene Blot-Verfahren, beschreibt den elektrophoretischen Transfer von Proteinen von SDS-Polyacrylamidgelen auf Nitrocellulosemembranen. Auch bekannt als Immunblotting-Protokoll.

View All Protocols




Global Trade Item Number

SKUGTIN
SNAP2RL04053252524455

Questions

Reviews

No rating value

Active Filters