About This Item
form
liquid
manufacturer/tradename
Specialty Media, EmbryoMax®
technique(s)
cell culture | stem cell: suitable
input
sample type neural stem cell(s)
sample type epithelial cells
sample type mesenchymal stem cell(s)
sample type hematopoietic stem cell(s)
sample type induced pluripotent stem cell(s)
sample type pancreatic stem cell(s)
sample type: mouse embryonic stem cell(s)
sample type cardiac stem cell(s)
sample type: human embryonic stem cell(s)
Preparation Note
Legal Information
Disclaimer
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
TrueGel3D® Hydrogel Plate protocol guides high-throughput culture of human adipose MSCs for screening applications.
Related Content
"As transgenic mouse core laboratories know well, mouse models are a powerful means of determining the function of mammalian genes in the context of whole organisms—especially since over 95% of the mouse genome is similar to the human genome. However, you may not know that: 1. Laboratory mouse strains are descended from ancestors in modern-day Pakistan. 2. The first inbred strain of lab mouse was created in 1909. It was named DBA for its coat color: dilute brown non-agouti. 3. There is a mutant mouse actually named“Bad Hair Day” (Bhrd). Someone get this little guy a stylist! 4. Male mice, when treated with anticancer drugs, have been shown to pass on DNA instability to their offspring. 5. The founder of Jackson Laboratories, C. C. Little, once had lunch with Walt Disney to request that an animated film be created linking Mickey Mouse and the laboratory mouse. The request was denied."
The cell culture protocols described in this instructional manual include the in vitro culture of murine ES cells using EmbryoMax products along with ESGRO® mLIF medium supplement, as well as feeder-free and serum-free ES cell culture using the ESGRO Complete™ line of products. Also included in this instructional manual are: methods for serum-free neuronal differentiation of mouse ES cells, iPS cell generation using STEMCCA™ lentivirus kits, cre-recombinase mediated excision of STEMCCA™ genes from iPS cells, derivation and rescue of new and existing ES cell lines using RESGRO™ Culture Medium and ESGRO Complete™ system.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| SLM-021-B | 04053252502859 |