Select a Size
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 1 mg | Check Cart for Availability | € 393,00 |
About This Item
€ 393,00
assay
≥98% (HPLC)
form
lyophilized
mol wt
calculated mol wt 1080.94
packaging
pkg of 1 mg
manufacturer/tradename
Millipore
storage condition
desiccated, protect from light
technique(s)
cell based assay: suitable
color
off-white
detection method
fluorometric
shipped in
ambient
storage temp.
−20°C
General description
BioTracker Rd1 Hypochlorite Live Cell Dye is a mitochondria-immobilized turn-on green fluorescent probe for the selective detection of hypochlorite (ClO-). The probe contains a benzoyl acetohydrazide linking site for ClO- recognition, a triphenylphosphonium group for mitochondria targeting, and a reactive methoxymaleimide subunit for mitochondrial fixation[1]. The targeting and fixed properties of the probe enable it to accumulate in mitochondria and prevents leakage of the dye upon the decrease of mitochondrial membrane potential (MMP). This probe may be used to visualize exogenous and endogenous ClO- levels in living cells, tissues, and zebrafish[1].
Reference:
1. Li, M. Y., et. al. (2020). Mitochondria-Immobilized Fluorescent Probe for the Detection of Hypochlorite in Living Cells, Tissues, and Zebrafishes. Analytical chemistry, 92(4), 3262-3269.
Application
Features and Benefits
Physical form
Preparation Note
Note: Centrifuge vial briefly to collect contents at bottom of vial before opening
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) cell based assay: suitable | technique(s) cell based assay: suitable | technique(s) cell based assay: suitable | technique(s) cell based assay: suitable, cell culture | stem cell: suitable, flow cytometry: suitable |
| color off-white | color yellow | color - | color - |
| form lyophilized | form lyophilized | form - | form - |
| storage temp. −20°C | storage temp. −20°C | storage temp. -10 to -25°C | storage temp. −20°C (−15°C to −25°C) |
| shipped in ambient | shipped in ambient | shipped in ambient | shipped in dry ice |
| assay ≥98% (HPLC) | assay ≥98% (HPLC) | assay ≥98% (H-NMR), ≥98% (HPLC), ≥98% (LC/MS), ≥98% (elemental analysis) | assay - |
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| SCT080 | 04061841827986 |



