Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|
About This Item
NACRES:
NA.45
UNSPSC Code:
41106200
Pricing and availability is not currently available.
General description
The EndoGRO® basal medium (Cat# SCME-BM) can be combined withfour different supplements to create four distinct media kits:
- To form EndoGRO®-LS Complete Media Kit use SCME001-S
- To form EndoGRO®-VEGFComplete Media Kit use SCME002-S
- To form EndoGRO®MV-VEGF Complete Media Kit use SCME003-S
- To form EndoGRO®-MV Complete Media Kit use SCME004-S
Application
EndoGRO® Media Formulations provide an optimal cell cultureenvironment for many types of endothelial cells, including HUVEC, aorticendothelial cells and other human large vessel and microvascular endothelialcells. These media have been shown to grow endothelial cells at rates that meetor exceed commercially available serum containing media, while maintainingexcellent cell morphology. The media are packaged in a specially designed UVprotective shrinkwrap for added stability, which includes a temperature gaugethat helps prevent contamination. EndoGRO® Media contain no antimicrobials andno phenol red, components that can cause cell stress and masking effects thatmay influence experimental results. Four different media formulations areavailable for various applications and cell culture requirements.
Preparation Note
Basal Medium (SCME-BM) should be stored at 2 - 8ºC. Thespecial UV protective packaging protects EndoGRO® Medium from light damage,however users should take care to protect medium from extended exposure tolight. Supplement Kits (SCME001-S, SCME002-S, SCME003-S, SCME004-S) should bestored at -20ºC. See individual labels for expiry date. Supplemented medium shouldbe stored at 2 - 8ºC in the dark. Use within 1 month of supplementation. Do notfreeze.
Other Notes
Unless otherwise stated in our catalog or other companydocumentation accompanying the product(s), our products are intended forresearch use only and are not to be used for any other purpose, which includesbut is not limited to, unauthorized commercial uses, in vitro diagnostic uses,ex vivo or in vivo therapeutic uses or any type of consumption or applicationto humans or animals.
Legal Information
ENDOGRO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
7791-20-0
CAS No.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| SCME-BM | 04053252610257 |