Skip to Content
Merck

Skip To

SCC602

3dGRO® Human Lung Organoids (LPTO.126)

Adenocarcinoma, Lung (Solid), Age: 81, Male, Mutation: Not Available

3dGRO® Human Lung Organoids (LPTO.126)

Synonym(s):

Lung PDO;Lung Tumor Organoid;Airway Organoid;Pulmonary Organoid, lung organoids

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
41106509
NACRES:
NA.28
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

biological source

human lung (solid)

packaging

vial of ≥ 1500 organoids/vial mg

manufacturer/tradename

Millipore

gender

male

growth mode

N/A

technique(s)

cell culture: suitable

application(s)

3D cell culture

control

lung (solid)

relevant disease(s)

adenocardinoma

shipped in

liquid nitrogen

storage temp.

−196°C

Application

  • Each vial contains >1500 organoids.
  • Mycoplasma: Negative
  • Genotyped by STR analysis: Pass
  • Human species verified
  • Organoids are negative for HPV-16, HPV-18, Hepatitis A, B, C, and HIV-1 & 2 viruses by PCR.
  • Cell viability test: Pass

Patient-derived lung organoids (lung PDOs) are adult tissue derived in vitro cultured “mini-lungs” that accurately replicate the histological and functional aspects of in vivo lung tissue. Lung organoids have been shown to contain all 6 major lung epithelial cell types including AEC2s, AEC1s, basal cells, club cells, goblet cells and ciliated cells. These 3D organoid models have become attractive alternatives to traditional PDX and animal lung models to study pulmonary diseases including COPD, cystic fibrosis, SARS-CoV-2, viral infections, asthma, and lung cancer. The 3dGRO® Human Lung Organoids are high quality cryopreserved cell lines derived from patient-derived xenografts, primary tumors or metastatic patient tissues from various regions of the human lung (acinar, papillary, solid, squamous, mucinous). These lung organoids are highly characterized and express lung airway markers including EPCAM, cytokeratin (CK5, CK7), acetyl-α-tubulin, surfactant proteins (SFTPB/SFTPC), TTF1, P63 and Muc5AC.

Cell Line Characteristics

Sex: Male
Age: 81
Organ: Lung (Primary Tumor- Solid)
Disease: Adenocarcinoma

Features and Benefits

Cryopreserved Male Adenocarcinoma Lung Organoids (Solid)

Preparation Note

3dGRO® human lung organoids should be stored in liquid nitrogen.

Legal Information

3dGRO® is a trademark of Merck KGaA, Darmstadt, Germany.
3dGRO® organoids were derived utilizing HUB Organoid Technology.

Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/pdouse. This product may not be re-engineered or copied; used to make derivatives or modifications; used to develop, test, or manufacture a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact [email protected].
3dGRO is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCC601SCC600SCC608
application(s)

3D cell culture

application(s)

3D cell culture

application(s)

3D cell culture

application(s)

3D cell culture

relevant disease(s)

adenocardinoma

relevant disease(s)

adenocardinoma

relevant disease(s)

adenocardinoma

relevant disease(s)

adenocardinoma

biological source

human lung (solid)

biological source

human lung (mucinous)

biological source

human lung (acinar)

biological source

human lung (Xenograph- Squamous)

technique(s)

cell culture: suitable

technique(s)

cell culture: suitable

technique(s)

cell culture: suitable

technique(s)

cell culture: suitable

storage temp.

−196°C

storage temp.

−196°C

storage temp.

−196°C

storage temp.

−196°C

packaging

vial of ≥ 1500 organoids/vial mg

packaging

vial of ≥ 1500 organoids/vial mg

packaging

vial of ≥ 1500 organoids/vial mg

packaging

vial of ≥ 1500 organoids/vial mg


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

Active Filters

  1. 1 Ratings-only review