Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
packaging
vial of >1X10⁶ cells
Quality Segment
manufacturer/tradename
Millipore
growth mode
N/A
technique(s)
cell culture | mammalian: suitable
shipped in
liquid nitrogen
storage temp.
−196°C
Application
- Each vial contains > 1X106 viable cells.
- Cells are tested negative for infectious diseases by a Human Essential CLEAR panel by Charles River Animal Diagnostic Services.
- Cells are verified to be of human origin and negative for inter-species contamination from mouse, rat, chinese hamster, Golden Syrian hamster, and Non-human Primate (NHP) as assessed by a Contamination Clear panel by Charles River Animal Diagnostic Services
- Cells are negative for mycoplasma contamination.
Adenocarcinoma presents a variety of challenges due to its resistance to common treatments such as radiotherapy and chemotherapy. Combining multiple forms of therapy helps to overcome these roadblocks. The Calu-3 cell line can assist in finding potential drug therapies and has shown to be useful in mimicking in vivo conditions.3 This similarity has allowed for many pharmaceutical applications such as drug transport, studying aerosol deposition, controlled release studies, and identification of potential drug-drug interactions.4 The characteristics of Calu3 cell line allow for a basic in vitro method of developing new therapeutics in an epithelial respiratory model.
Source
The Calu-3 cell line was established by isolating cells located in the pleural effusion of a 25-year-old Caucasian man with lung adenocarcinoma.5
References
1. Semin Ultrasound CT MR 2019, 40(3): 255-264.
2. Cell Death Dis 2018, 9(2): 117.
3. J Control Release 2003, 87(1-3): 131-138.
4. Expert Opin Drug Deliv 2013, 10(9): 1287-1302.
5. J Natl Cancer Inst 1977, 59(1): 221-226.
Features and Benefits
Preparation Note
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable | technique(s) cell culture | mammalian: suitable |
| growth mode N/A | growth mode N/A | growth mode N/A | growth mode N/A |
| shipped in liquid nitrogen | shipped in liquid nitrogen | shipped in liquid nitrogen | shipped in liquid nitrogen |
| storage temp. −196°C | storage temp. −196°C | storage temp. −196°C | storage temp. −196°C |
| manufacturer/tradename Millipore | manufacturer/tradename Millipore | manufacturer/tradename Millipore | manufacturer/tradename Millipore |
| packaging vial of >1X10⁶ cells | packaging vial of >1X10⁶ cells | packaging vial of >1X10⁶ cells | packaging vial of >1X10⁶ cells |
Storage Class
10 - Combustible liquids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| SCC438 | 04065267115528 |



