Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Pack Size | SKU | Availability | Price |
|---|---|---|---|
| 1 vial | Check Cart for Availability | € 1.050,00 |
About This Item
NACRES:
NA.81
UNSPSC Code:
41106514
€ 1.050,00
Check Cart for Availability
application(s)
cell analysis
General description
Squamous cell carcinoma of the head and neck (HNSCC) is the 9th-most common cancer worldwide and is characterized by a high rate of recurrence after therapy, with a median 5-year survival range of 40-50% (1). HNSCC tumors show a wide range of heterogeneity in response to radiation therapy. Cellular HNSCC models that demonstrate a range of radiation sensitivity are thus invaluable for elucidating the factors underlying radiation responses of cancer cells.
The SCC-61 HNSCC cell line, derived from an aggressive tongue squamous cell carcinoma (2), is an established model for HNSCC. SCC-61 cells are relatively sensitive to radiation compared to other HNSCC cell lines as well as a relatively stable genome (3). SCC-61 cells grow robustly and are strongly adherent in culture. SCC-61 cells have been characterized by positive expression of the SCC marker p63 (p40) (4). SCC-61 cells have demonstrated utility in studies parsing mechanisms of radio-resistance in SCC tumor cells, as demonstrated by transformation of this cell line with factors conferring radio-resistance (5).
<bold>Source:</bold>
SCC-61 cell line was derived from a tongue squamous cell carcinoma of a male patient (2).
Research Category:
Cancer
The SCC-61 HNSCC cell line, derived from an aggressive tongue squamous cell carcinoma (2), is an established model for HNSCC. SCC-61 cells are relatively sensitive to radiation compared to other HNSCC cell lines as well as a relatively stable genome (3). SCC-61 cells grow robustly and are strongly adherent in culture. SCC-61 cells have been characterized by positive expression of the SCC marker p63 (p40) (4). SCC-61 cells have demonstrated utility in studies parsing mechanisms of radio-resistance in SCC tumor cells, as demonstrated by transformation of this cell line with factors conferring radio-resistance (5).
<bold>Source:</bold>
SCC-61 cell line was derived from a tongue squamous cell carcinoma of a male patient (2).
Research Category:
Cancer
Biochem/physiol Actions
Human, Cancer Cells
Packaging
≥1X106 cells/vial
Preparation Note
Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.
Other Notes
Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/restrictedcelluse. This product may not be: re-engineered or copied; used to make derivatives, modifications or functional equivalents; used to obtain patents or other IP claiming use of the product; used to develop, test, or manufacturer a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact [email protected].
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
1 of 1
This Item | |||
|---|---|---|---|
| application(s) cell analysis | application(s) cell analysis | application(s) - | application(s) - |
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| SCC280 | 04061842104062 |



