Skip to Content
Merck

Skip To

SCC077

UM-SCC-22B Squamous Carcinoma Cell Line

Human

Synonym(s):

UMSCC22A

HNSCC

Head and neck squamous cell carcinoma cell line

H&N squamous cell carcinoma cell line

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
41106514
NACRES:
NA.75
eCl@ss:
32011203
Biological source:
human
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

UM-SCC-22B Squamous Carcinoma Cell Line, UM-SCC-22B is a unique head and neck squamous carcinoma cell line established from the metastatic lymph node in the neck of a female patient and is a suitable in vitro model of H&N carcinoma studies.

biological source

human

Quality Segment

technique(s)

cell culture | mammalian: suitable

shipped in

liquid nitrogen

storage temp.

-140 to -196°C

General description

All UM-SCC cell lines were derived in the lab of Dr. Thomas Carey at the University of Michigan and are exclusively distributed by Merck KGaA, Darmstadt, Germany. Access to UM-SCC cells is limited solely to those officers, employees and students of PURCHASER’s institution who need access to perform research. PURCHASER may not distribute UM-SCC cells or derivatives to third parties. PURCHASER shall comply with all applicable laws in its use and handling of the Product and shall keep it under reasonable safe and secure conditions to prevent unauthorized use or access.

THIS PRODUCT IS ONLY AVAILABLE FOR SALE TO ACADEMIC INSTITUTIONS OR NOT-FOR-PROFIT ENTITIES FOR USE UNDER THIS LIMITED USE LABEL LICENSE. FOR INFORMATION ON COMMERCIAL LICENSING OF UM-SCC CELLS, INCLUDING LICENSING TO COMMERCIAL ENTITIES, PLEASE CONTACT: [email protected]
UM-SCC-22B is a unique head and neck squamous carcinoma cell line established from the metastatic lymph node in the neck of a female patient [1] [2]. UM-SCC-22B is negative for HPV-16, HPV-18, Hepatitis A, B, C, and HIV-1 & 2 viruses.

UM-SCC-22B was established at the University of Michigan with written informed consent obtained from the patient and with the approval of the study by the Medical School Institutional Review Board .

Application

Subject to local law, this product is intended to be sold for internal in vitro research use only subject to terms and conditions found here: www.sigmaaldrich.com/restrictedcelluse. This product may not be: re-engineered or copied; used to make derivatives, modifications or functional equivalents; used to obtain patents or other IP claiming use of the product; used to develop, test, or manufacturer a commercial product; used as a component in a commercial product; resold or licensed; used in any clinical applications or trials; or used in humans. A license or limited commercial use agreement is required for use by any for-profit entity, use in services, and use in sponsored academic research. For information regarding any such use, please contact [email protected].

Biochem/physiol Actions

Cancer Cells

Preparation Note

UM-SCC-22B cells should be stored in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Analysis Note

• Each vial contains ≥ 1X106 viable cells.
• Cells are tested by PCR and are negative for HPV-16, HPV-18, Hepatitis A, B, C and HIV-1 & 2 viruses.
• Cells are negative for mycoplasma contamination.
• Each lot of cells are genotyped by STR analysis to verify the unique identity of the cell line.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SCC076SCC073SCC074
biological source

human

biological source

human

biological source

human

biological source

human

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

technique(s)

cell culture | mammalian: suitable

storage temp.

-140 to -196°C

storage temp.

-140 to -196°C

storage temp.

-140 to -196°C

storage temp.

-140 to -196°C

shipped in

liquid nitrogen

shipped in

dry ice

shipped in

liquid nitrogen

shipped in

dry ice

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Zhenkun Zhu et al.
Molecular pharmaceutics, 7(4), 1283-1290 (2010-06-15)
The small heat shock protein 27 (Hsp27) is a molecular chaperone that is involved in a variety of cellular functions in cancer cells. The purpose of this research is to study Hsp27 in vitro metastatic behaviors of head and neck
J Chad Brenner et al.
Head & neck, 32(4), 417-426 (2009-09-18)
We established multiple University of Michigan Squamous Cell Carcinoma (UM-SCC) cell lines. With time, these have been distributed to other labs all over the world. Recent scientific discussions have noted the need to confirm the origin and identity of cell



Global Trade Item Number

SKUGTIN
SCC07704055977333497

Questions

Reviews

No rating value

Active Filters