Skip to Content
Merck
All Photos(1)

Key Documents

S7117

Sigma-Aldrich

ApopTag Plastic Coverslip

APOPTAG Plastic Coverslips are designed to be custom fitted to the specific APOPTAG kit application.

Synonym(s):

Plastic coverslip

Sign Into View Organizational & Contract Pricing

Select a Size

1000 EA
€630.00

€630.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
1000 EA
€630.00

About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.32

€630.00


Estimated to ship on24 May 2025


Request a Bulk Order

Quality Level

manufacturer/tradename

ApopTag
Chemicon®

technique(s)

activity assay: suitable (apoptosis)

shipped in

ambient

General description

APOPTAG Plastic Coverslips are designed to be custom fitted to the specific APOPTAG kit application. Each coverslip has 2.5" x 1" dimensions.

Application

Plastic coverslips can be used to assure that a constant volume of solution is applied per unit of specimen area. However, their handling time slows down the protocol. Each square centimeter of plastic coverslip will require the volume of reagent indicated in Table 5 (S7100, S7101) and Table 10 (S7110, S7111, S7160), so that the reagent volume applied per unit of tissue area can be held constant. The surface to be covered is always equal to the area of the plastic coverslip, not the area of the specimen.

Plastic coverslips may be trimmed to any desired size and shape. The kit′s yield of specimens will be reduced if larger than standard coverslips are used.

When using ApopTag Kits that contain the anti-digoxigenin antibodies (S7100, S7101, S7110, S7111), plastic coverslips can be used during the incubation steps with the following reagents: WORKING STRENGTH TdT, and the ANTI-DIGOXIGENIN ANTIBODY. When using the ApopTag Fluorescein Direct Kit (S7160), the plastic coverslips can be used during incubation steps with WORKING STRENGTH TdT.

A basic coverslip method is described as follows:

To make a pair of "standard area" (~5 cm2) specimen coverslips, cut a plastic coverslip (provided) into two equal halves, and fold up a 1 cm handling tab across the width, then crease sharply (See Figure 3 in insert).

Drain one slide for approximately 10 seconds, and then tap off drops on a paper towel on the benchtop. Blot back and sides of the slide with a folded wipe. Carefully blot the area around the tissue section or cells, or else vacuum up solution using a pipette attached to an aspirator vacuum.

Apply reagent solution to one end of the area to be covered, using a dropper bottle or pipette as required.

Grasp the plastic coverslip by the handling tab and touch its opposite end to the droplet of reagent on the slide. Slightly arching the coverslip, roll it slowly downward, causing the solution to spread by capillary action. If solution does not spread evenly, tilt the slide until the flow reaches all edges.

Apply plastic coverslips to microscope slides so as to minimize trapped air bubbles, which may cause variable enzyme reaction or detection.

Place the slide across the pipettes, face-up and level, inside the humidified chamber. The slide edges should not touch anything so as to prevent drainage of the reagent.

Storage and Stability

Store at room temperature.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service