Skip to Content
Merck

Skip To

R03465

Lactophenol

mounting fluid for demonstrating microscopic morphology of fungi

Synonym(s):

Fungi Stain

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
41116130


Quality Segment

color

blue

application(s)

diagnostic assay manufacturing
hematology
histology

shipped in

ambient

storage temp.

15-25°C

General description

Mounting fluid for demonstrating microscopic morphology of fungi

Application

Mounting fluid for demonstrating microscopic morphology of fungi
Prepared from dyes certified by the Biological Stain Commission (BSC)
For In-Vitro Diagnostic Use.
For Professional Use.
Mounting fluid for demonstrating microscopic morphology of fungi.


signalword

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Muta. 2 - Skin Corr. 1B - STOT RE 2

target_organs

Nervous system,Kidney,Liver,Skin

supp_hazards

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Related Content

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

Ein Überblick über die Wissenschaft und Praxis der Bakteriologie in der klinischen Diagnostik. Informationen über die Anwendung von Standard- und Spezialfärbungen für die mikroskopische Analyse.





Questions

Reviews

No rating value

Active Filters