Key Documents
PUMP05001
Milli-Q
Recirculation Pump
AC/DC input 230 V
About This Item
UNSPSC Code:
41104200
NACRES:
SB.51
Pricing and availability is not currently available.
AC/DC input
230 V, 50-60 Hz
packaging
pkg of 1 unit
compatibility
for use with tanks
General description
Recirculation Pump is a water purification system accessory. The pump recirculates stored water to maintain pure water quality.
For use with 30, 60, and 100 L polyethylene (PE) reservoirs to protect stored water quality.