Skip to Content
Merck

Skip To

69959

Perfect Protein Western Markers

Accurate size markers detectable on any Western blot

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 assays
Check Cart for Availability
€ 106,00

About This Item

NACRES:
NC.01
UNSPSC Code:
41105337

€ 106,00


Check Cart for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

manufacturer/tradename

Novagen®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−20°C

General description

The Perfect Protein<TMSYMBOL></TMSYMBOL> Western Markers consist of seven recombinant S•Tag<TMSYMBOL></TMSYMBOL>/His•Tag<TMSYMBOL></TMSYMBOL> fusion proteins that can be detected on any Western blot using an S-protein AP or HRP conjugate. As an additional option, the markers can also be detected with an His•Tag Monoclonal Antibody and an AP or HRP conjugated secondary antibody. The Perfect Protein Western Markers serve as precise size standards that appear simultaneously on the blot with target proteins, eliminating the uncertainty and imprecision associated with other methods.

For ease of convenience, please consider the Perfect Protein AP Western Blot Kit, or the Perfect Protein HRP Western Blot Kit. For a version containing prestained proteins for tracking electrophoresis and Western transfer, please refer to our complete listing of Trail Mix Western Markers and Blot Kits.

A.Two parallel blots were incubated with primary antibodies for an expressed protein, then incubated with Anti-Mouse IgG AP Conjugate and processed using chemiluminescent detection. The S-protein AP Conjugate was included with secondary antibody for the blot on the left. Lanes: 1, 5 l Perfect Protein Western Markers; 2, 10 ng induced cell extract; 3, 1 g uninduced cell extract.
B.Blot contains the Perfect Protein Western Markers plus purified GUS expressed in insect cells incubated with a GUS-specific monoclonal antibody, then incubated with Anti-Mouse IgG HRP Conjugate and S-protein HRP Conjugate and detected with chemiluminescent HRP substrate. Lanes: 1, 5 l Perfect Protein Western Markers; 2, 800 ng purified GUS protein.

Features and Benefits

• Detect markers simply by adding the S-protein(AP or HRP Conjugate) or His•Tag MonoclonalAntibody to the same incubation used forsample detection.

• S-protein conjugate (included in kits) doesnot interfere with antibodies or streptavidindetection.

• Can be used with colorimetric andchemiluminescent AP or HRP substrates.

• Recombinant, unmodified markers give sharp,accurately sized bands.

• Markers are supplied at the working dilution ingel loading buffer; concentration optimized forWestern detection.

• Protein sizes are 15, 25, 35, 50, 75, 100, and150 kDa.

Legal Information

NOVAGEN is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Toxic (F)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
6907969149-M2504
form

liquid

form

liquid

form

liquid

form

liquid

shipped in

wet ice

shipped in

wet ice

shipped in

-

shipped in

wet ice

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

storage condition

-

manufacturer/tradename

Novagen®

manufacturer/tradename

Novagen®

manufacturer/tradename

Novagen®

manufacturer/tradename

Chemicon®, Re-Blot


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters