About This Item
biological source
rabbit
Quality Level
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
liquid
contains
≤0.1% sodium azide as preservative
species reactivity
human
manufacturer/tradename
Calbiochem®
storage condition
OK to freeze
dilution
(Immunoblotting (1:1000)
ELISA (1:2000))
isotype
IgG
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
phosphorylation (pSer188)
Gene Information
human ... RHOA(387)
General description
Immunogen
Application

Immunoblotting (1:1000)
ELISA (1:2000)
Packaging
Physical form
Preparation Note
Analysis Note
RhoA GST fusion protein
Other Notes
Sawada, N., et al. 2001. Biochem. Biophys. Res. Commun. 280, 798.
Dong, J.M., et al. 1998. J. Biol. Chem.273, 22554.
Lang, P., et al. 1996. EMBO J.15, 510.
Legal Information
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in wet ice |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| Gene Information human ... RHOA(387) | Gene Information human ... CHEK1(1111) | Gene Information human ... MAPK14(1432) | Gene Information human ... TP53(7157) |
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
WGK 1
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| PS1089-100UL | 04055977226577 |



