Skip to Content
Merck

Skip To

PS1023

PhosphoDetect Anti-Smad3 (pSer423/425) Rabbit mAb (C25A9)

liquid, clone C25A9, Calbiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
50 μL
Check Cart for Availability
€ 335,00

About This Item

NACRES:
NA.43
UNSPSC Code:
12352203
Clone:
C25A9, monoclonal
Species reactivity:
human
Application:
Citations:
7

€ 335,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

C25A9, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

human

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

dilution

(Immunoblotting (1:1000)
Immunoprecipitation (1:50))

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer423/pSer425)

Gene Information

human ... SMAD3(4088)

General description

Purified rabbit monoclonal antibody generated by immunizing rabbits with the specified immunogen and fusing splenocytes with an immortal cell line. Recognizes the ~52 kDa Smad3 protein phosphorylated at Ser423/425.
Recognizes the ~52 kDa Smad3 protein phosphorylated at Ser423/425 in HT-1080 cells treated with TGF-β1.
This PhosphoDetect Anti-Smad3 (pSer423/425) Rabbit mAb (C25A9) is validated for use in Immunoblotting, Immunoprecipitation for the detection of Smad3 (pSer423/425).

Immunogen

a synthetic phosphopeptide corresponding to amino acids surrounding the Ser 423/425 phosphorylation sites of Smad3, coupled to KLH.

Application


Immunoblotting (1:1000)
Immunoprecipitation (1:50)

Physical form

In 150 mM NaCl, 10 mM HEPES, 100 µg/ml BSA, 50% glycerol pH 7.5.

Other Notes

Attisano, L., et al. 2002. Science 296, 1646.
Moustakas, A., et al. 2001. J. Cell Sci. 114, 4359.
Wrana, J. 2000. Science 23, 1.
Does not cross-react with other Smad proteins. For immunoblotting incubate the membrane with antibody diluted with TBS containing 5% BSA, 0.1% Tween®-20 detergent overnight at 4°C with gentle shaking. Please note that this is a rabbit monoclonal antibody and requires the use of an anti-rabbit secondary antibody. The immunogen sequence is 100% conserved in mouse, rat, monkey, bovine, xenopus, and zebrafish Smad3, but cross-reactivity has not been tested. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AP1063361527PK1010
biological source

rabbit

biological source

rabbit

biological source

mouse

biological source

rabbit

antibody form

purified antibody

antibody form

affinity isolated antibody

antibody form

purified antibody

antibody form

affinity isolated antibody

species reactivity

human

species reactivity

mouse, human, rat

species reactivity

mouse, human

species reactivity

human, mouse

Gene Information

human ... SMAD3(4088)

Gene Information

-

Gene Information

human ... GSK3B(2932)
mouse ... Gsk3B(56637)

Gene Information

human ... SYK(6850)

clone

C25A9, monoclonal

clone

polyclonal

clone

2D3, monoclonal

clone

polyclonal

form

liquid

form

liquid

form

liquid

form

liquid


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Amy M Gancarz et al.
Nature neuroscience, 18(7), 959-961 (2015-06-02)
Activin receptor signaling, including the transcription factor Smad3, was upregulated in the rat nucleus accumbens (NAc) shell following withdrawal from cocaine. Direct genetic and pharmacological manipulations of this pathway bidirectionally altered cocaine seeking while governing morphological plasticity in NAc neurons.
Javier Milara et al.
Respiratory research, 23(1), 313-313 (2022-11-16)
Pulmonary hypertension (PH) associated to idiopathic pulmonary fibrosis (IPF) portends a poor prognosis. IL-11 has been implicated in fibrotic diseases, but their role on pulmonary vessels is unknown. Here we analyzed the contribution of IL-11 to PH in patients with
Beatriz Ballester et al.
International journal of molecular sciences, 22(12) (2021-07-03)
Several transmembrane mucins have demonstrated that they contribute intracellularly to induce fibrotic processes. The extracellular domain of MUC16 is considered as a biomarker for disease progression and death in IPF patients. However, there is no evidence regarding the signalling capabilities



Global Trade Item Number

SKUGTIN
PS1023-50UL04055977227116

Questions

Reviews

No rating value

Active Filters