Select a Size
| Pack Size | SKU | Availability | Price |
|---|
About This Item
Product Name
Ultracel® 30 kDa Ultrafiltration Discs, Chemistry: Regenerated Cellulose (RC) Filter Diameter: 90 mm Amicon® Bioseparations
material
regenerated cellulose membrane
Quality Level
feature
hydrophilic
manufacturer/tradename
Ultracel®
technique(s)
protein clean-up: suitable (desalting), protein purification: suitable (concentration)
diam.
90 mm
filter diam.
90 mm
filtration area
63 cm2
matrix
Ultracel®
pore size
30 kDa NMWCO
shipped in
ambient
General description
Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic, tight microstructure of Ultracel membranes assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.
Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.
Microfiltration Durapore PVDF cut discs are available 0.1 µm, 0.22 µm, and 0.45 µm pore sizes in both 63.5 mm and 75 mm diameters.
Features & Benefits:
•Disc diameters include: 25, 44.5, 47, 63.5, 76, 90 and 150 mm
•Both Ultracel, Durapore, and Biomax membranes are available in a wide range of pore sizes to meet your separation requirements
•Compatible with Amicon Stirred Cells
Applications:
Concentration, Buffer Exchange, Desalting
Application
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename Ultracel® | manufacturer/tradename Ultracel® | manufacturer/tradename Ultracel® | manufacturer/tradename Ultracel® |
| material regenerated cellulose membrane | material regenerated cellulose membrane | material regenerated cellulose membrane | material regenerated cellulose membrane |
| diam. 90 mm | diam. 44.5 mm | diam. 47 mm | diam. 76 mm |
| pore size 30 kDa NMWCO | pore size 30 kDa NMWCO | pore size 30 kDa NMWCO | pore size 30 kDa NMWCO |
| feature hydrophilic | feature hydrophilic | feature hydrophilic | feature hydrophilic |
| shipped in ambient | shipped in ambient | shipped in ambient | shipped in ambient |
Still not finding the right product?
Explore all of our products under Ultracel® 30 kDa Ultrafiltration Discs
Storage Class
11 - Combustible Solids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
The best in large-volume ultrafiltration just got better. Introducing the new Amicon® Stirred Cell. You’re an Amicon® Stirred Cell owner. You’re adept at finely separating macrosolutes in large-volume (50-400 mL) samples, and you depend on the precision of Amicon® devices. Or maybe you are an expert in membrane analysis, and you count on a device that lets you insert membranes of your choice. With your needs in mind, EMD Millipore has developed the new Amicon® Stirred Cell, providing the same gentle, high recovery of macrosolutes and thorough buffer exchange to which you’re well accustomed. It provides the same membrane flexibility and ability to monitor filtration progress.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| PLTK09005 | 04053252613203 |

