Skip to Content
Merck

Skip To

PHCCCABLE

Scepter USB Cable

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
41104923
NACRES:
NB.22
eCl@ss:
32011202


manufacturer/tradename

Scepter

technique(s)

cell counting: suitable

shipped in

ambient

storage temp.

no temp limit

Application

Research Category
Cell Culture

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EP542885028078080BCMT12130716
storage temp.

no temp limit

storage temp.

-

storage temp.

room temp

storage temp.

-

shipped in

ambient

shipped in

-

shipped in

-

shipped in

-

technique(s)

cell counting: suitable

technique(s)

-

technique(s)

-

technique(s)

-

manufacturer/tradename

Scepter

manufacturer/tradename

5428850280, Eppendorf®

manufacturer/tradename

MVP ICON®

manufacturer/tradename

Mettler-Toledo 12130716


Storage Class

10-13 - German Storage Class 10 to 13



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library


Protocols

Combine the ease of automated instrumentation with the accuracy of impedance-based cell counting using the Coulter principle in an affordable, handheld format.

The Scepter™ cell counter uses the Coulter principle of impedance-based particle detection to reliably and accurately count every cell in your sample.





Global Trade Item Number

SKUGTIN
PHCCCABLE04053252552175

Questions

Reviews

No rating value

Active Filters