Sign In to View Organizational & Contract Pricing.
About This Item
NACRES:
NA.43
UNSPSC Code:
12352203
Clone:
polyclonal
Species reactivity:
human, mouse
Application:
—
Citations:
11
biological source
rabbit
Quality Level
antibody form
purified antibody
antibody product type
primary antibodies
clone
polyclonal
form
liquid
contains
≤0.1% sodium azide as preservative
species reactivity
human, mouse
manufacturer/tradename
Calbiochem®
storage condition
do not freeze
isotype
IgG
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
human ... MLH1(4292)
General description
Purified rabbit polyclonal antibody. Recognizes the ~83 kDa MLH1 protein.
Recognizes the ~83 kDa MLH1 protein.
This Anti-MLH1 (Ab-2) Rabbit pAb is validated for use in Immunoblotting for the detection of MLH1 (Ab-2).
Immunogen
Human
recombinant human MLH1
Application
Immunoblotting (1 µg/ml)
Packaging
Please refer to vial label for lot-specific concentration.
Physical form
In 0.05 M sodium phosphate buffer, 0.2% gelatin.
Analysis Note
Positive Control
LoVo cells
LoVo cells
Negative Control
HCT116 cells
HCT116 cells
Other Notes
Antibody should be titrated for optimal results in individual systems.
Bronner, C.E., et al. 1994. Nature368, 258.
Papadopoulos, N., et al. 1994. Science263, 1625.
Peltomäki, P.T. 1994. Ann. Med.26, 215.
Fishel, R., et al. 1993. Cell75, 1027.
Leach, F.S., et al. 1993. Cell75, 1215.
Lindbolm, A., et al. 1993. Nat. Genet.5, 279.
Papadopoulos, N., et al. 1994. Science263, 1625.
Peltomäki, P.T. 1994. Ann. Med.26, 215.
Fishel, R., et al. 1993. Cell75, 1027.
Leach, F.S., et al. 1993. Cell75, 1215.
Lindbolm, A., et al. 1993. Nat. Genet.5, 279.
Legal Information
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Toxicity: Standard Handling (A)
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
10 - Combustible liquids
wgk
nwg
flash_point_f
Not applicable
flash_point_c
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Tegan S Horan et al.
PLoS genetics, 13(7), e1006885-e1006885 (2017-07-21)
The hypothesis that developmental estrogenic exposure induces a constellation of male reproductive tract abnormalities is supported by experimental and human evidence. Experimental data also suggest that some induced effects persist in descendants of exposed males. These multi- and transgenerational effects
Martha Susiarjo et al.
Methods in molecular biology (Clifton, N.J.), 558, 339-354 (2009-08-18)
The goal of this chapter is twofold: First, to acquaint the reader with the problems inherent in analyzing mammalian female meiosis and, second, to provide a step-by-step approach to mastering the necessary techniques. Although the methods presented are for use
Lisa A Vrooman et al.
Genetics, 196(2), 385-396 (2013-12-10)
Increasing age in a woman is a well-documented risk factor for meiotic errors, but the effect of paternal age is less clear. Although it is generally agreed that spermatogenesis declines with age, the mechanisms that account for this remain unclear.
Global Trade Item Number
| SKU | GTIN |
|---|---|
| PC56-100UG | 04055977207798 |