Skip to Content
Merck

Skip To

PC150

Anti-β-Amyloid (1-42) Rabbit pAb

liquid, Calbiochem®

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
25 μg
Check Cart for Availability
€ 474,00

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Clone:
polyclonal
Species reactivity:
human
Application:
Citations:
3

€ 474,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

does not contain

preservative

species reactivity

human

packaging

antibody small pack of 25 μg

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, avoid repeated freeze/thaw cycles

dilution

(Dot Blot (0.1-0.5 µg/mL)
ELISA (0.1-0.5 µg/mL)
Radioimmunoassay (0.5 µg/mL))

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... APP(351)

General description

Rabbit polyclonal antibody adsorbed against Aβ1-40 and Aβ1-43, followed by immunoaffinity purification. Recognizes specifically β-amyloid peptide (1-42).
Recognizes specifically β-amyloid peptide (1-42). Does not cross-react with β-amyloid peptide (1-40) or β-amyloid peptide (1-43).

  • Antibody Target Gene Symbol: APP
  • Target Synonym: AAA, AD1, Adap, AL024401, Amyloid precursor, Amyloidogenic glycoprotein, APP isoform 1, APPI, appican, CTFgamma, CVAP, E030013M08RIK, Nexin II, P3, PN2, PreA4, PROTEASE NEXIN2
  • Entrez Gene Name: amyloid beta (A4) precursor protein
  • Hu Entrez ID: 351 (Related Antibodies: 171610, PC729,PC152, PC151, PC149, NE1012, 171609, 171608, 171607, 171606, 171605, 171604, 171603, 171598, 171537)
  • Mu Entrez ID: 11820
  • Rat Entrez ID: 54226
This Anti-β-Amyloid (1-42) Rabbit pAb is validated for use in Dot Blot, ELISA, Radioimmunoassay for the detection of β-Amyloid (1-42).

Immunogen

Human
a synthetic peptide corresponding to amino acids from a C-terminal fragment of human β-amyloid peptide (1-42)

Application

Dot Blot (0.1-0.5 µg/ml)

ELISA (0.1-0.5 µg/ml)

Radioimmunoassay (0.5 µg/ml)

Packaging

Please refer to vial label for lot-specific concentration.

Physical form

In phosphate buffer, pH 7.4.

Preparation Note

Following initial thaw, aliquot and freeze (-20°C).

Other Notes

Savage, M.J., et al. 1998. J. Neurosci.15, 1743.
Borchelt, D.R., et al. 1997. Neuron19, 939.
Citron, M., et al. 1997. Nature Med.3, 67.
Hardy, J. 1997. Trends in Neurosci.20, 154.
Solomon, B., et al. 1997. Proc. Natl. Acad. Sci. USA94, 4109.
Duff, K., et al. 1996. Nature383, 710.
Iwatsubo, T., et al. 1994. Neuron13, 45.
Suzuki, N., et al. 1994. Science264, 133.
This antibody is specific for β-amyloid peptide [1-42] and shows no reactivity to β-amyloid peptide [1-40] or β-amyloid peptide [1-43]. This antibody is recommended for use in the quantitative determination of β-amyloid peptide [1-42]. The sensitivity level for sandwich ELISA is 15-25 pg. Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Standard Handling (A)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
ABN240AB5078PAB5074P
Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity purified immunoglobulin

antibody form

affinity purified immunoglobulin

biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

form

liquid

form

-

form

-

form

-

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

dry ice

storage temp.

−20°C

storage temp.

-

storage temp.

-

storage temp.

-


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
PC150-25UG04055977209471

Questions

Reviews

No rating value

Active Filters