Skip to Content
Merck

Skip To

NM0045

NovaSeal Battery Powered Crimping Tool

AC/DC input 220 V / 240 V AC, UK plug, Kit content : crimping tool, 10,8V Li-ion battery, battery charger

Synonym(s):

NovaSeal Electrical Crimping Tool

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

eCl@ss:
32010310
UNSPSC Code:
41121815
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Level

sterility

non-sterile

product line

NovaSeal

compatibility

for use with NovaSeal pinch pipes


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
NM0043NM0044A100-M
product line

NovaSeal

product line

NovaSeal

product line

NovaSeal

product line

NovaSeptum®

compatibility

for use with NovaSeal pinch pipes

compatibility

for use with NovaSeal pinch pipes

compatibility

for use with NovaSeal pinch pipes

compatibility

for use with NovaSeptum® tube disconnection pinch pipes. NS6-15-05PK100

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

sterility

non-sterile

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

SP2


Still not finding the right product?


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
NM004504054839079306

Questions

Reviews

No rating value

Active Filters