Skip to Content
Merck

Skip To

NB-2ML-B

Blood typing BIOSCOT IVD Reagent Other Specificities & AHG

BIOSCOT Anti-Lea from the clone GA2, Murine IgA, Blood Grouping Reagent

Synonym(s):

Anti-Lea (clone GA2), Blood Grouping Reagent

Sign In to View Organizational & Contract Pricing.

About This Item

NACRES:
NA.41
UNSPSC Code:
41116102
Clone:
GA2, monoclonal
Technique(s):
agglutination assay: suitable
Application:
Agglut.
Citations:
-
Note: For further information on price, availability or to place an order, please contact us


biological source

mouse

Quality Level

clone

GA2, monoclonal

packaging

pkg of 2 mL

manufacturer/tradename

BIOSCOT

storage condition

do not freeze

concentration

3-12 mg/mL protein (Total Protein Range)

technique(s)

agglutination assay: suitable

pH

6-8

isotype

IgA

application(s)

clinical testing

shipped in

ambient

storage temp.

2-8°C

General description

Contains 0.1% (w/v) Sodium Azide

Filtered through a 0.22μm terminal filter

Shelf Life: 36 months from date of filtration

CE marked under the requirements of EU regulation 2017/746


For In Vitro use only

Application

For use by Tube technique
Vialled, Labeled

Legal Information

BIOSCOT is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
NB-1L-BKNB-10X2ML-BNB-10X2ML-U
biological source

mouse

biological source

mouse

biological source

mouse

biological source

mouse

clone

GA2, monoclonal

clone

GA2, monoclonal

clone

GA2, monoclonal

clone

GA2, monoclonal

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

technique(s)

agglutination assay: suitable

technique(s)

agglutination assay: suitable

technique(s)

agglutination assay: suitable

technique(s)

agglutination assay: suitable

Quality Level

600

Quality Level

600

Quality Level

600

Quality Level

600

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient


Still not finding the right product?


Storage Class

10 - Combustible liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library





Global Trade Item Number

SKUGTIN
NB-2ML-B04053252707353