Merck
Skip To
MXQUA4VP2
Milliflex® Quantum system Validation Protocol for the combination with Milliflex® Oasis system
for use with Milliflex® Quantum Systems, for use with Milliflex Oasis® Systems
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
Size/SKUAvailabilityPrice
About This Item
NACRES:
ND.05
UNSPSC Code:
41116500
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
availability
not available in North America
compatibility
for use with Milliflex Oasis® Systems, for use with Milliflex® Quantum Systems
General description
Milliflex® Quantum Validation Protocol(A4 Format) is designed as a ready to use step by step guide to perform IQ and OQ activities of rapid microbial detection system using Milliflex® Quantum in combination with Milliflex® Oasis system. The protocol contains an exhaustive list of tests including test descriptions and test records. The protocol follows GMP, EP, USP and JP regulatory guidelines. Not available in North America.
Application
Milliflex® Quantum Validation Protocol is used to validate your rapid microbial detection system with ease and consistent performance while following regulatory guidelines.
Legal Information
MILLIFLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
MILLIFLEX OASIS is a registered trademark of Merck KGaA, Darmstadt, Germany
Compare Similar Items
View Full Comparison
Show Differences
1 of 1
This Item
MXQULTVP2RPSTA4VP1MMQUANK01
compatibility
for use with Milliflex Oasis® Systems, for use with Milliflex® Quantum Systems
compatibility
for use with Milliflex Oasis® Systems, for use with Milliflex® Quantum Systems
compatibility
for use with Milliflex® Rapid Systems.
compatibility
for use with Milliflex® Quantum system.
availability
not available in North America
availability
-
availability
-
availability
-