Skip to Content
Merck

Skip To

MX1583A

Molecular Sieve, Type 3A

pellet diam. 1/16 in.

Synonym(s):

Molecular Sieve

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

UNSPSC Code:
23201100
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

pellets, rod

pellet diam.

1/16 in.

color

tan

shipped in

ambient

storage temp.

room temp

General description

1/16 Pellets

Application


  • Study on the Adsorption of Trace Water in N-Methyl-pyrrolidone Solvents by A-Type Molecular Sieves: This research explores the use of type 3A molecular sieves for the adsorption of trace water in N-Methyl-pyrrolidone solvents, demonstrating their effectiveness in solvent dehydration processes (Zhang J et al., 2024).

Analysis Note

Color: Tan
Diameter: 1/16 inchinch
Form: Pellets/rods
Identification (potassium): To pass test


Still not finding the right product?

Explore all of our products under Molecular Sieve, Type 3A


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class

8B - Non-combustible corrosive hazardous materials

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters