Skip to Content
Merck

Skip To

MX1458

Dimethyl sulfoxide

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Synonym(s):

Dimethyl sulfoxide, Dimethyl Sulfoxide, DMSO, DMSO

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

Linear Formula:
(CH3)2SO
CAS Number:
Molecular Weight:
78.13
EC Index Number:
200-664-3
UNSPSC Code:
12352119
Beilstein/REAXYS Number:
506008
MDL number:
Assay:
≥99.9% (GC)
Bp:
189 °C (lit.)
Vapor pressure:
0.42 mmHg ( 20 °C)
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


agency

meets USP/NF

Quality Segment

vapor density

2.7 (vs air)

vapor pressure

0.42 mmHg ( 20 °C)

assay

≥99.9% (GC)

form

liquid

autoignition temp.

573 °F

expl. lim.

42 %, 63 °F

dilution

(for analytical testing)

impurities

≤0.001 meq/g Titrable acid, ≤0.1% Water (H2O)

evapn. residue

≤0.01%

color

APHA: ≤10

refractive index

n20/D 1.479 (lit.)

bp

189 °C (lit.)

mp

16-19 °C (lit.)

transition temp

flash point 87 °C

solubility

1000 g/L

density

1.10 g/mL (lit.)

shipped in

ambient

storage temp.

room temp

SMILES string

CS(C)=O

InChI

1S/C2H6OS/c1-4(2)3/h1-2H3

InChI key

IAZDPXIOMUYVGZ-UHFFFAOYSA-N

Application

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs GR ACS

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

AppearanceClear, colorless liquidAssay (GC)99.9% minColor (APHA)10 maxEvaporation residue0.01% maxTitrable acid0.001meq/g maxWater (H2O)0.1% max

Disclaimer

Supercools easily and remelts slowly at room temperature. Solidified product can be re-liquified by warming to room temperature without detriment to the product.


Still not finding the right product?

Explore all of our products under Dimethyl sulfoxide


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

188.6 °F - closed cup

flash_point_c

87 °C - closed cup



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
MX1458-3104061839430457
MX1458-604061839430525
MX1458-304061839430433
MX1458-504061839430501

Questions

Reviews

No rating value

Active Filters