Skip to Content
Merck

Skip To

MAC132

Mouse Anti-Human IgA, Secretory Antibody, Clone 4G10.1

clone 4G10.1, from mouse

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice
400 μg
Check Cart for Availability
€ 299,00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.46
eCl@ss:
32160702
Clone:
4G10.1, monoclonal
Species reactivity:
human (IgA)
Application:
ELISA
Citations:
-

€ 299,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

mouse

Quality Segment

antibody form

purified immunoglobulin

antibody product type

secondary antibodies

clone

4G10.1, monoclonal

species reactivity

human (IgA)

technique(s)

ELISA: suitable

isotype

IgG1κ

shipped in

wet ice

target post-translational modification

unmodified

General description

The Mouse Anti-Human IgA antibody is an IgG monomeric immunoglobulin, built of two heavy chains and two light chains. Each molecule has two antigen binding sites. This is the most abundant immunoglobulin and is approximately equally distributed in blood and in tissue liquids, constituting 75% of serum immunoglobulins in humans. This is the only isotype that can pass through the human placenta, thereby providing protection to the fetus in its first weeks of life before its own immune system has developed. It can bind to many kinds of pathogens, for example viruses, bacteria, and fungi, and protects the body against them by complement activation (classic pathway), opsonization for phagocytosis and neutralisation of their toxins. There are 4 subclasses: IgG1 (66%), IgG2 (23%), IgG3 (7%) and IgG4 (4%). It is affinity-purified and demonstrated to react with Human IgA with minimal or no cross-reaction to Human IgG or Human IgM. The target of this antibody, Human IgA, constitutes 5-15 % of the serum immunoglobulins whereas dimeric IgA is localized to mucosa surfaces such as saliva, gastrointestinal secretion, bronchial fluids and milk. Mucosal IgA plays a major role in host defence by neutralising infectious agents at mucosal surfaces. The production is usually local and antigen specific IgA producing B-cells can be found in regions under the lamina propria where they mature into dimeric IgA producing plasma cells. IgA deficiency is the most common immunodeficiency that may affect both serum and mucosal produced IgA.

Immunogen

Purified antibody corresponding to Human IgA, Secretory.

Application

ELISA. Can be used as a capture antibody for ELISA at 10 μg/ml in PBS without carrier protein. Antibody should be titrated for optimal results in individual systems.
Research Category
Secondary & Control Antibodies
Research Sub Category
Secondary Antibodies Adsorbed for Dual Labeling
This Mouse Anti-Human IgA, Secretory Antibody, Clone 4G10.1 is validated for use in Enzyme Immunoassay (ELISA) for the detection of Mouse Human IgA, Secretory.

Biochem/physiol Actions

Based on ELISA analysis, this antibody recognizes human IgA secretory subunit, with minimal or no cross-reaction human IgG or IgM.

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing PBS without preservatives.

Preparation Note

Stable for 1 year at -20°C from date of receipt.

Analysis Note

Evaluated by ELISA against Human IgA, Human IgG and Human IgM

Other Notes

Concentration: Please refer to lot specific datasheet.
Replaces: 411423-400UG

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
AP501AQ502AQ502A
biological source

mouse

biological source

goat

biological source

goat

biological source

goat

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

technique(s)

ELISA: suitable

antibody form

purified immunoglobulin

antibody form

affinity purified immunoglobulin

antibody form

purified antibody

antibody form

purified antibody

species reactivity

human (IgA)

species reactivity

mouse

species reactivity

mouse

species reactivity

mouse

isotype

IgG1κ

isotype

IgA, IgG, IgM

isotype

-

isotype

-

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice

shipped in

wet ice


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Global Trade Item Number

SKUGTIN
MAC13204053252978838

Questions

Reviews

No rating value

Active Filters