Skip to Content
Merck

Skip To

MABT820

Anti-Integrin alpha 5 (ITGA5/CD49E) Antibody

rat monoclonal, mAb16

Synonym(s):

CD49 antigen-like family member E, Fibronectin receptor subunit alpha, Integrin alpha-F, VLA-5, CD49e

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Pack SizeSKUAvailabilityPrice

About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
Clone:
mAb16, monoclonal
Species reactivity:
human
Application:
ICC, IP, inhibition assay
Citations:
4
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Anti-Integrin alpha 5 (CD49e) Antibody, clone mAb16, clone mAb16, from rat

biological source

rat

Quality Segment

antibody form

purified antibody

antibody product type

primary antibodies

clone

mAb16, monoclonal

species reactivity

human

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable, inhibition assay: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

ambient

target post-translational modification

unmodified

Gene Information

human ... ITGA5(3678)

General description

114.54 kDa calculated.
Integrin alpha-5 (UniProt: P08648; also known as CD49 antigen-like family member E, Fibronectin receptor subunit alpha, Integrin alpha-F, VLA-5, CD49e) is encoded by the ITGA5 (also known as FNRA) gene (Gene ID: 3678) in human. Integrins are heterodimeric integral membrane proteins composed of an alpha subunit and a beta subunit that function in cell surface adhesion and signaling. Integrin alpha-5 is a single-pass type I membrane protein is expressed on multiple cell types. It contains four calcium binding sites (aa 480-488; 334-342; 401-409; and 465-473). Structurally, it consists of an extracellular domain (aa 4-995), a transmembrane domain (aa 996-1021), and a cytoplasmic domain (aa 1022-1049). When associated with beta 1 subunit it can serve as a receptor for fibronectin and fibrinogen. It recognizes the sequence R-G-D in its ligands. Integrin alpha-5 is also expressed in articular cartilage and plays a role in synovial joint development and in the regulation of chondrocyte survival and matrix degradation in articular cartilage. Membrane fractions obtained from fibroblasts of diabetic animals display significant reduction in integrin alpha-5 levels. (Ref.: Almeida, ME et al., (2016). Mol. Cell. Biochem. 421(1-2); 19-28).

Immunogen

Fibronectin receptor purified from human placenta.

Application

Anti-Integrin alpha5 (CD49e), clone mAb16, Cat. No. MABT820, is a rat monoclonal antibody that detects Integrin alpha5 and has been tested for use in Immunocytochemistry, Immunoprecipitation, and Inhibition assays.
Inhibition Analysis: 10 ug/mL from a representative lot prevented HT-1080 cells from adhering to fibronectin coated plates (Courtesy of Dr. Kenneth M. Yamada, M.D., Ph.D., NIDCR, National Institutes of Health, Bethesda, MD USA).

Inhibition Analysis: A representative lot inhibited WI38 fibroblast attachment to fibronectin (Akiyama, S.K., et. al. (1989). J Cell Biol. 109(2):863-75).

Inhibition Analysis: A representative lot inhibited A375-SM cell spreading. (Humphries, J.D., et. al. (2005). J Biol Chem. 280(11):10234-43).

Immunocytochemistry Analysis: A representative lot detected Integrin alpha5 (CD49e) in Immunocytochemistry applications (Akiyama, S.K., et. al. (1989). J Cell Biol. 109(2):863-75).


Immunoprecipitation Analysis: A representative lot immunoprecipitated Integrin alpha5 (CD49e) in Immunoprecipitation applications (Akiyama, S.K., et. al. (1989). J Cell Biol. 109(2):863-75).
Research Category
Cell Structure

Biochem/physiol Actions

Clone mAb16 is a rat monoclonal antibody that specifically detects Integrin alpha5 in human cells.

Physical form

Format: Purified
Protein G purified
Purified rat monoclonal antibody IgG2a in PBS without azide.

Preparation Note

Stable for 1 year at -20°C from date of receipt.

Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.

Analysis Note

Evaluated by Immunocytochemistry in HFF cells.

Immunocytochemistry Analysis: A 1:25 dilution of this antibody detected Integrin alpha5 (CD49e) in human Foreskin Fibroblast (HFF) cells.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
MAB1956ZMABT822MABT207
biological source

rat

biological source

mouse

biological source

rat

biological source

mouse

Gene Information

human ... ITGA5(3678)

Gene Information

human ... ITGA5(3678)

Gene Information

human ... ITGA5(3678)
mouse ... Itga5(16402)

Gene Information

human ... ITGAV(3685)

antibody form

purified antibody

antibody form

purified antibody

antibody form

purified immunoglobulin

antibody form

purified antibody

species reactivity

human

species reactivity

human

species reactivity

human, mouse

species reactivity

human

clone

mAb16, monoclonal

clone

P1D6, monoclonal

clone

mAB11, monoclonal

clone

272-17E6, monoclonal

technique(s)

immunocytochemistry: suitable, inhibition assay: suitable, immunoprecipitation (IP): suitable

technique(s)

immunohistochemistry: suitable

technique(s)

immunocytochemistry: suitable, immunoprecipitation (IP): suitable

technique(s)

activity assay: suitable, flow cytometry: suitable, immunoprecipitation (IP): suitable


Still not finding the right product?

or

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library







Global Trade Item Number

SKUGTIN
MABT82004054839416583

Questions

Reviews

No rating value

Active Filters