Skip to Content
Merck
All Photos(1)

Key Documents

MABT1550

Sigma-Aldrich

Anti-Lactadherin/MFGE8 Antibody, clone 5C5

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
€146.00
100 μL
€369.00

€146.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
25 μL
€146.00
100 μL
€369.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€146.00


Estimated to ship on25 May 2025


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

5C5, monoclonal

mol wt

calculated mol wt 45.73 kDa
observed mol wt ~47 kDa

purified by

using protein G

species reactivity

human, boar

packaging

antibody small pack of 100 μL

technique(s)

immunofluorescence: suitable
western blot: suitable

isotype

IgG1κ

epitope sequence

Unknown

UniProt accession no.

shipped in

2-8°C

target post-translational modification

unmodified

Gene Information

General description

Lactadherin (UniProt: P79385; also known as MFGM, Milk fat globule-EGF factor 8, MFG-E8, SED1, Sperm surface protein SP47, PP47) is encoded by the MFGE8 gene (Gene ID: 397545) in Boar. MFG-E8 is a peripheral membrane glycoprotein that is widely expressed in many cell types. At its N-terminal region, it contains two EGF-like repeats (aa 2-41 and 44-88). The second EGF repeat contains an RGF integrin-binding domain that allows its interaction with and activation of Vβ3 and Vβ5 integrins. It also has two C-terminal F5/8 type C domains. The second F5/8 type C domain exhibits high affinity for binding to phosphatidylserine present in cell membranes. MFG-E8 displays high affinity for glycoproteins on egg surface and contributes to the ability of sperm to adhere to the egg in an integrin-independent manner. It has been reported that although MFG-E8 is not absolutely essential for fertilization, its absence is shown to reduce fertility in mice. It has also been implicated in a variety of other physiological functions. MFG-E8 can bind apoptotic cells via its F5/8C domains, and its RGD motif is shown to be essential for cell engulfment by macrophages. In the mammary glands, it is required for clearance of apoptotic cells during mammary gland involution at the end of lactation. Defects in or lack of MFG-E8 can cause the accumulation of apoptotic cell remnants, duct distension and mastitis. (Ref.: Zigo, M., et al. (2015). Cell Tissue Res. 359(3); 895-908; Ensslin, MA., and Shur, BD. (2007). Proc. Natl. Acad. Sci. USA 104(8); 2715-2720).

Specificity

Clone 5C5 is a mouse monoclonal antibody that detects Sperm surface protein (Lactadherin).

Immunogen

Purified Sperm surface protein from Boar.

Application

Quality Control Testing

Evaluated by Western Blotting in MDA-MB-231 cell lysate.

Western Blotting Analysis: A 1:500 dilution of this antibody detected Lactadherin/MFGE8 in MDA-MB-231 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:500 dilution from a representative lot detected Lactadherin/MFGE8 in HeLa cell lysate.

Western Blotting Analysis: A representative lot detected Lactadherin/MFGE8 in Western Blotting applications (Zigo, M., et al. (2015). Cell Tissue Res. 359(3):895-908).

Immunofluorescence Analysis: A representative lot detected Lactadherin/MFGE8 in Immunofluorescence applications (Zigo, M., et al. (2015). Cell Tissue Res. 359(3):895-908).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-Lactadherin/MFGE8 , clone 5C5, Cat. No. MABT1550, is a mouse monoclonal antibody that detects Lactadherin and is tested for use in Immunofluorescence and Western Blotting.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Recommend storage at +2°C to +8°C. For long term storage antibodies can be kept at -20°C. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service