Skip to Content
Merck
All Photos(1)

Key Documents

914940

Sigma-Aldrich

Cerium Zirconium Oxide Milled Nanofiber (Ce:Zr ratio 50:50)

Synonym(s):

CeZrO4 nanofiber, Nanofibrous inorganic powder structure

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€164.00
10 G
€283.00

€164.00


0 left in stock

Request a Bulk Order

Select a Size

Change View
5 G
€164.00
10 G
€283.00

About This Item

Linear Formula:
CeZr04
CAS Number:
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23

€164.00


0 left in stock

Request a Bulk Order

form

nanofiber

Quality Level

fiber diam. × L

300-800 nm × 210 μm , Structure: Polycystalline nanofibers

surface area

30-60 m2/g

primary crystallite size

(tetragonal
Typical Size of Crystallites: 8 nm)

SMILES string

[Ce].[Zr]=O

InChI

1S/Ce.O.Zr

InChI key

GJFXIYGDVYZDRX-UHFFFAOYSA-N

Application

Cerium Zirconium oxide has applications in automotive combustion sensor and controller for catalytic convertors, catalyst support for hot gas desulfurization, fuel cells, catalyst and catalyst support, thermochemical water splitting, infrared filters, and inorganic separators.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Review discusses recent progress and challenges in ORR catalyst development for PEMFC applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service